Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
13-247-725F
Fisher Scientific™ Isotemp™ Premium Lab Ovens: Forced Air
Full-feature ovens available in a variety of sizes
Includes:
  • Small oven supplied with one shelf (holds up to 5)
  • Medium oven has one shelf (holds up to 8)
  • Large oven comes with two shelves (holds up to 11)
Operating Features
  • Intuitive microprocessor controls with touch keypad, LED digital display and on-screen prompts
  • Variable temperature control from 50° to 275°C in 1°C increments
  • Automatic overtemperature protection at 5°C above setpoint with visual alarm
  • Last settings retained in memory
  • Built-in safety backup and circuit breaker to protect oven from power surges
Cabinet Features
  • Easy-to-clean stainless-steel chambers
  • Enamel-painted steel exterior with coated aluminum trim
  • Low-watt-density, longlife heating elements
  • Thick 21/2 in. (6.4cm) all-around fiberglass insulation keeps exterior cool
  • Gasketed door has stay-cool latch; opens 180° for easy access
  • Adjustable chrome-plated steel wire shelves (163/4 x 173/8 in. [44 x 42.5cm])
  • Nonskid rubber feet elevate cabinet for ventilation
Three Sizes
Small: 2.5 cu. ft. chamber volume; supplied with one shelf (holds up to 5)
Medium: 3.75 cu. ft.; with one shelf (holds up to 8)
Large: 5.0 cu. ft.; with two shelves (holds up to 11) Forced-Air Ovens
  • Deliver more uniform heating for better temperature control and faster drying
  • Blower circulates air throughout chamber; fresh air is mixed in and recirculated
  • Used air vented through top of oven
View Specifications
Images for Fisher Scientific™ Isotemp™ Premium Lab Ovens: Forced Air
  • 13-247-725F
13-247-725F
13-247-725F
(5 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13-247-738F Medium, Forced Air; 240V
Accessories
Each for $3,698.38
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
13-247-726F Small, Forced Air, 240V
Accessories
N/A N/A N/A
This product is regulated by the federal government, and may not be ordered online. 
13-247-725F Small; Forced Air; 120V
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
13-247-750F Large, Forced Air; 120V
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
13-247-737F Medium; Forced Air; 120V
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
Operating Features
  • Intuitive microprocessor controls with touch keypad, LED digital display and on-screen prompts
  • Variable temperature control from 50° to 275°C in 1°C increments
  • Automatic overtemperature protection at 5°C above setpoint with visual alarm
  • Last settings retained in memory
  • Built-in safety backup and circuit breaker to protect oven from power surges
Cabinet Features
  • Easy-to-clean stainless-steel chambers
  • Enamel-painted steel exterior with coated aluminum trim
  • Low-watt-density, longlife heating elements
  • Thick 21/2 in. (6.4cm) all-around fiberglass insulation keeps exterior cool
  • Gasketed door has stay-cool latch; opens 180° for easy access
  • Adjustable chrome-plated steel wire shelves (163/4 x 173/8 in. [44 x 42.5cm])
  • Nonskid rubber feet elevate cabinet for ventilation
Three Sizes
Small: 2.5 cu. ft. chamber volume; supplied with one shelf (holds up to 5)
Medium: 3.75 cu. ft.; with one shelf (holds up to 8)
Large: 5.0 cu. ft.; with two shelves (holds up to 11) Forced-Air Ovens
  • Deliver more uniform heating for better temperature control and faster drying
  • Blower circulates air throughout chamber; fresh air is mixed in and recirculated
  • Used air vented through top of oven
Specifications & Ordering Information:
Shelf 13-247S is available online
Warranty: 12 months
1Average valve performed per ASTM* E 145.
2Door open 1 minute at 200°C.
3Length includes thickness of door; does not include handle.
4Units require a 20-amp outlet (NEMA 5–20R).