Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Thermo Scientific™ Thermolyne™ Largest Tabletop Muffle Furnaces
1.6 cu. ft. (45.3L) capacity furnaces with choice of programmable or single-setpoint controller
Includes: Two refractory shelves
Required Accessories: Electrical connection requires fixed wiring (no power cord).
  • Advanced LED digital-set/digital-display temperature controller is microprocessor-controlled
  • LED display simultaneously shows both setpoint and actual furnace temperatures in °C or °F
  • User-selectable overtemperature protection Open thermocouple protection
  • Adjustable power output from 1 to 100%
Safety and Design Features
  • Heating elements are on chamber top, bottom and sides for enhanced temperature uniformity
  • Built-in vent port removes undesirable contaminants and moisture to extend the life of the element and unit
  • Rear of chamber incorporates a 0.95cm (0.38 in.) diameter port for monitoring chamber temperatures with independent measuring devices
  • Critical electronic components and heating elements are protected by a 35A circuit breaker
  • Door safety switch stops power to the heating elements when door opens
View Specifications
Images for Thermo Scientific™ Thermolyne™ Largest Tabletop Muffle Furnaces
(5 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
10-505-10
Thermo Scientific
No.:F30420C
Single Setpoint; 240V 22.9A
10-505-13
Thermo Scientific
No.:F30428C
Single Setpoint; 208V 26.4A
10-505-14
Thermo Scientific
No.:F30438CM
Multiprogrammable; 208V 26.4A
10-505-62
Thermo Scientific
No.:F30430CM
Multiprogrammable; 240V 22.9A
NC9755732
Thermo Scientific
No.:USED/F30420C-80/PRC
Programmable, 240V, 22.9A
Description
  • Advanced LED digital-set/digital-display temperature controller is microprocessor-controlled
  • LED display simultaneously shows both setpoint and actual furnace temperatures in °C or °F
  • User-selectable overtemperature protection Open thermocouple protection
  • Adjustable power output from 1 to 100%
Safety and Design Features
  • Heating elements are on chamber top, bottom and sides for enhanced temperature uniformity
  • Built-in vent port removes undesirable contaminants and moisture to extend the life of the element and unit
  • Rear of chamber incorporates a 0.95cm (0.38 in.) diameter port for monitoring chamber temperatures with independent measuring devices
  • Critical electronic components and heating elements are protected by a 35A circuit breaker
  • Door safety switch stops power to the heating elements when door opens
Applications:
  • Annealing glass
  • Determinations of volatiles
  • Catalyst research
  • Ashing organic and inorganic samples
Choice of Temperature Controllers
  • Furnaces that use B1, C1 or D1 control also use a mechanical Overtemperature Protection relay
  • B1: Digital single setpoint control with a single ramp to setpoint and dwell; single display shows actual temperature or setpoint
  • C1: Digital programmable control with one stored program of 8 segments
  • D1: Digital programmable control with 4 stored programs, 16 segments per program, and RS-232 communications interface
Certifications: All units CSA approved; -33 units also CE marked.
Specifications
Temperature Range100° to 1093°C
Temperature Stability±1.2°C at 1000°C
Temperature Uniformity±3.45°C
Capacity 45L (1.6 cu. ft.)
Interior D x W x H 36 x 36 x 36cm (14 x 14 x 14 in.)
Exterior L x W x H 64.7 x 54.6 x 74.9cm (25.5 x 21.5 x 29.5 in.)
Shipping Weight117.9kg (260 lb.)
Power Consumption5500w