Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
07-200-567, 07-200-568
Available on GSA/VA Contract for Federal Government customers only. Costar™ 96-Well Black or White Clear-Bottom Plates
For fluorescent and luminescent assays; ideal for performing cell-based and molecular assays
  • Molded polystyrene with opaque walls and optically clear bottoms
  • Flat 6.4mm diameter wells
  • Surface area per well: 0.32cm2
  • Available in either solid black or white wall colors
  • Choose from untreated, high-binding or tissue culture-treated plates
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Costar™ 96-Well Black or White Clear-Bottom Plates
  • 07-200-567, 07-200-568
07-200-567, 07-200-568
07-200-567, 07-200-568
(7 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
07-200-565 Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Plastic
No.:3603
Tissue culture treated; Black
Case of 48 for $397.48
Check Availability
Add
to Cart
(0) in My Cart
07-200-566 Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Plastic
No.:3610
Tissue culture treated; White
Case of 48 for $401.50
Check Availability
Add
to Cart
(0) in My Cart
07-200-567 Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Plastic
No.:3631
Untreated; Black
Case of 100 for $662.00
Check Availability
Add
to Cart
(0) in My Cart
07-200-568 Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Plastic
No.:3632
Untreated; White
Case of 100 for $694.50
Check Availability
Add
to Cart
(0) in My Cart
07-200-587 Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Plastic
No.:3903
Tissue Culture treated; White
Case of 100 for $786.50
Check Availability
Add
to Cart
(0) in My Cart
07-200-588 Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Plastic
No.:3904
Tissue culture treated; Black
Case of 100 for $733.65
Check Availability
Add
to Cart
(0) in My Cart
07-200-625 Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Plastic
No.:3601
High binding; Black
Case of 100 for $778.00
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Molded polystyrene with opaque walls and optically clear bottoms
  • Flat 6.4mm diameter wells
  • Surface area per well: 0.32cm2
  • Available in either solid black or white wall colors
  • Choose from untreated, high-binding or tissue culture-treated plates
Specifications & Ordering Information:
Cat. No. 07-200-568 also available in a pack of 25.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.