Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
18-999-625
Replacement Smart Sensors for Honeywell Analytics™ PhD™ Ultra Gas Detectors
  • The detectors must be recalibrated before being returned to service following any sensor change
View Specifications
Images for Replacement Smart Sensors for Honeywell Analytics™ PhD™ Ultra Gas Detectors
  • 18-999-625
18-999-625
18-999-625
(3 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
18-999-625
Biosystems
No.:54-04-90
Type: oxygen (O2)
Each for $205.00
Check Availability
Add
to Cart
(0) in My Cart
18-999-628
Biosystems
No.:54-04-02
Type: hydrogen sulfide (H2S)
Each for $307.50
Check Availability
Add
to Cart
(0) in My Cart
18-999-627
Biosystems
No.:54-04-01
Type: carbon monoxide (CO)
Each for $307.50
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • The detectors must be recalibrated before being returned to service following any sensor change