Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
22-274-209
View Image Gallery (1) Image(s) are representative of the product Group
  • 22-274-209
Fisher Scientific* Midget Urinometer
Accuracy guaranteed to ±0.002
  • Set consists of a glass jar and standard shot ballast hydrometer with a smooth outside wall
  • Range of measurements is from 1.000 to 1.040 in 0.001 increments
View Specifications
Images for Fisher Scientific* Midget Urinometer
  • 22-274-209
22-274-209
(3 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Midget urinometer set; 1-1.04x.001
22-274-209 N/AYou must be logged in and have a valid account number to view item availability   Each for $42.48
Case of 12 EA for $429.22
(0) in My Cart
Replacement hydrometer midgut; Measurement range: 1-1.040
22-274-217 N/AYou must be logged in and have a valid account number to view item availability   Each for $29.84
Case of 12 EA for $301.58
(0) in My Cart
Jar Urinometer 12/cs
22-274-225 N/AYou must be logged in and have a valid account number to view item availability   Each for $5.20
Case of 12 EA for $62.43
(0) in My Cart
Add Items to Cart
Description
  • Set consists of a glass jar and standard shot ballast hydrometer with a smooth outside wall
  • Range of measurements is from 1.000 to 1.040 in 0.001 increments