Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Fisherbrand™ 6 in. (150mm) Ruler
With cork and stopper size guide
  • Blue vinyl
  • Dual-scale ruler; 0.062 in. and 1mm increments
  • Temperature conversion scale on back for –20° to +100°C and –10° to +212°F ranges
View Specifications
Images for Fisherbrand™ 6 in. (150mm) Ruler
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
09-016 Length: 6 in. (150mm)
Pack of 100 for $88.00
Pack of 100 for $66.00 Online Instant Savings (25%)
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Blue vinyl
  • Dual-scale ruler; 0.062 in. and 1mm increments
  • Temperature conversion scale on back for –20° to +100°C and –10° to +212°F ranges