Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
14-906 Series
View Image Gallery (6) Image(s) are representative of the product Group
  • 14-906 Series
  • 1490627
  • 1490628
  • 1490631
  • 1490629
  • 1490630
Available on GSA/VA Contract for Federal Government customers only. Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack
Calibrated 1 and 10µL plastic disposable inoculating loops and needles
Zip-lock closure reseals the pack between use to avoid contamination. Re-sealable bag allows the user to open and close the loop pack to remove individual loops.
  • Disposable, sterile plastic loops and needles manufactured from soft-flexible or hard-rigid plastic
  • Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
  • Ultrasmooth surfaces allow easy streaking without cutting or gouging agar surface
  • Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
  • Color coded for easy product identification
  • Packed in tamper-proof bags to guarantee sterility
  • Certificate of calibration included in each case
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack
  • 14-906 Series
  • 1490627
  • 1490628
  • 1490631
  • 1490629
  • 1490630
14-906 Series
14-906 Series
(5 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
14-906-27 Available on GSA/VA Contract for Federal Government customers only. 
Copan Diagnostics
No.:COPH1
Rigid; 1µL; Dark green
Case of 1000 for $178.92
Case of 1000 for $152.08 Online Instant Savings (15%)
Check Availability
Add
to Cart
(0) in My Cart
Alternative Product(s)
Show Alternative
Also available: Fisherbrand™ Disposable Inoculating Loops and Needles , which is an alternative option to catalog number  1490627
Ideal for use in laboratories Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and
Catalog No. Description Price Per Unit Quantity & Availability Add Item(s)
22363604
Volume: 1µL; Color: Blue Pack of 960 for $175.71
Case of 10 PK for $1,580.97
Check Availability
Add to
Cart
14-906-28 Available on GSA/VA Contract for Federal Government customers only. 
Copan Diagnostics
No.:COP-H10
Rigid; 10µL; Dark blue
Case of 1000 for $178.92
Case of 1000 for $152.08 Online Instant Savings (15%)
Check Availability
Add
to Cart
(0) in My Cart
Alternative Product(s)
Show Alternative
Also available: Fisherbrand™ Loops and Needles , which is an alternative option to catalog number  1490628
No need to flame metal loops and expose to airborne pathogens   ·  Disposable, sterile plastic loops and needles for quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation  ·  Molded from biologically inert
Catalog No. Description Price Per Unit Quantity & Availability Add Item(s)
22363599
Rigid Loop; Volume: 10µL, Light Blue Case of 1000 for $218.35
Check Availability
Add to
Cart
Show Alternative
Also available: Fisherbrand™ Disposable Inoculating Loops and Needles , which is an alternative option to catalog number  1490628
Ideal for use in laboratories Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and
Catalog No. Description Price Per Unit Quantity & Availability Add Item(s)
22363606
Volume: 10µL, Color: Yellow; 1000Pk Case of 1000 for $184.14
Check Availability
Add to
Cart
14-906-29 Available on GSA/VA Contract for Federal Government customers only. 
Copan Diagnostics
No.:COPS1
Flexible; 1µL; Light green
Case of 1000 for $178.92
Case of 1000 for $152.08 Online Instant Savings (15%)
Check Availability
Add
to Cart
(0) in My Cart
Alternative Product(s)
Show Alternative
Also available: Fisherbrand™ Loops and Needles , which is an alternative option to catalog number  1490629
No need to flame metal loops and expose to airborne pathogens   ·  Disposable, sterile plastic loops and needles for quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation  ·  Molded from biologically inert
Catalog No. Description Price Per Unit Quantity & Availability Add Item(s)
22363595
Flexible Loop; Volume: 1µL; Color: Light blue Pack of 960 for $301.94
Case of 10 PK for $2,683.27
Check Availability
Add to
Cart
Show Alternative
Also available: Fisherbrand™ Disposable Inoculating Loops and Needles , which is an alternative option to catalog number  1490629
Ideal for use in laboratories Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and
Catalog No. Description Price Per Unit Quantity & Availability Add Item(s)
22363604
Volume: 1µL; Color: Blue Pack of 960 for $175.71
Case of 10 PK for $1,580.97
Check Availability
Add to
Cart
14-906-30 Available on GSA/VA Contract for Federal Government customers only. 
Copan Diagnostics
No.:COPS10
Flexible; 10µL; Light blue
Case of 1000 for $178.92
Case of 1000 for $152.08 Online Instant Savings (15%)
Check Availability
Add
to Cart
(0) in My Cart
Alternative Product(s)
Show Alternative
Also available: Fisherbrand™ Loops and Needles , which is an alternative option to catalog number  1490630
No need to flame metal loops and expose to airborne pathogens   ·  Disposable, sterile plastic loops and needles for quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation  ·  Molded from biologically inert
Catalog No. Description Price Per Unit Quantity & Availability Add Item(s)
22363597
Flexible Loop; Volume: 10µL; Color: Yellow Pack of 960 for $301.61
Case of 10 PK for $2,679.72
Check Availability
Add to
Cart
14-906-31 Available on GSA/VA Contract for Federal Government customers only. 
Copan Diagnostics
No.:COPNED
1.45mm diameter;20cm length; Violet
Case of 1000 for $178.92
Case of 1000 for $152.08 Online Instant Savings (15%)
Check Availability
Add
to Cart
(0) in My Cart
Alternative Product(s)
Show Alternative
Also available: Fisherbrand™ Loops and Needles , which is an alternative option to catalog number  1490631
No need to flame metal loops and expose to airborne pathogens   ·  Disposable, sterile plastic loops and needles for quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation  ·  Molded from biologically inert
Catalog No. Description Price Per Unit Quantity & Availability Add Item(s)
22363596
Needle; Color: White Pack of 960 for $280.13
Case of 10 PK for $2,330.30
Check Availability
Add to
Cart
Add Items to Cart
Description
  • Disposable, sterile plastic loops and needles manufactured from soft-flexible or hard-rigid plastic
  • Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
  • Ultrasmooth surfaces allow easy streaking without cutting or gouging agar surface
  • Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
  • Color coded for easy product identification
  • Packed in tamper-proof bags to guarantee sterility
  • Certificate of calibration included in each case
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.