Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S40802
Fisher Scientific™ Traceable™ Digital Barometer
Ideal for recording ambient conditions in the lab, monitoring differences that could affect sample analysis, and predicting weather changes
Combination unit simultaneously displays current barometric pressure, bar graph of barometric trends, humidity, temperature, and date or time. Switches permit setting in °F or °C, and  in.Hg or mbar. Use toggle switch to display the month and date.
Includes: Lab-bench stand, wall mount, and four “AAA” batteries.
  • Electronic digital barometer has memory feature that automatically captures reading every hour—press HISTORY button to display the last 24 readings
  • Bar graph shows current reading and the trend for the previous 1, 3, 6, 12, and 24 hours
  • Visual indicator displays weather forecast of sunny, cloudy, or rainy
View Specifications
Images for Fisher Scientific™ Traceable™ Digital Barometer
  • S40802
S40802
S40802
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
02-400 Digital barometer
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
  • Electronic digital barometer has memory feature that automatically captures reading every hour—press HISTORY button to display the last 24 readings
  • Bar graph shows current reading and the trend for the previous 1, 3, 6, 12, and 24 hours
  • Visual indicator displays weather forecast of sunny, cloudy, or rainy
Specifications & Ordering Information:
Measures 1L x 5.25W x 7.25 in.H (2.5 x 13 x 18cm). Net weight: 10 oz. (283.5g)
Certifications: NIST traceable
Compliance: Traceable to standards provided by NIST™. Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA.
Specifications
Atmospheric Pressure
Range23.45 to 31.01 in. Hg/794 to 1050mbar
Resolution0.03 in. Hg (1mbar)
Accuracy±0.1477 in. Hg/±5mbar
Altitude Compensation0 to 2500m (For atmp. pressure)
Temperature
Range32° to 130°F/0° to +55°C
Resolution0.90°F/0.50°C
Accuracy±1°C/±1.8°F
ResponseUpdates every 9 seconds
Relative Humidity
Range10 to 98% RH
Resolution1%
Accuracy±5 RH
ResponseUpdates every 9 seconds
TimeA clock displays time to the minute in either AM/PM or military time