Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Fisher Scientific™ Isotemp™ Refrigerator and Freezer Alarm Systems
For Isotemp and Isotemp Plus™ refrigerators and freezers
Required Accessories: Remote systems require an existing alarm system, with dry alarm contacts, to be installed on the refrigerator or freezer in order to be compatible.
Freestanding Alarm System
  • Use with basic freezers, refrigerators, and one section on refrigerator/freezer combinations With key switch in ALARM position, system provides audio and visual alarms if chamber temperature reaches high or low alarm limit (adjustable, set by user) or if power is lost for more than 30 seconds
  • Housed in a painted metal cabinet with digital temperature display
  • Alarm silencing and test features
  • Can be used to trigger Standard or Deluxe Remote Alarm systems
Standard Remote System†
  • Use with refrigerators and freezers equipped with panel-mounted or freestanding alarm (Alarm or Recorder/Alarm models)
  • Can be located up to one-third mile from unit
  • Emits audible and visual signals in the event of power failure, temperature deviation, or if door is open for more than three minutes
  • Alarm sounds if line between unit and alarm is cut
Deluxe Remote System†
  • Use with refrigerators and freezers equipped with panel-mounted or freestanding alarm (Alarm or Recorder/Alarm models)
  • Can monitor up to four individual or four groups of refrigerators or freezers
  • User-programmable to call up to eight telephone or pager numbers when activated by power failure, temperature deviation, or if door is open for more than three minutes
  • Alarm sounds if line between unit and alarm is cut
  • Stores four outgoing messages
  • Automatic redial for busy or unanswered numbers
  • Works with any standard telephone system
  • Battery backup allows alarm to sound in the event of a power failure
View Specifications
Images for Fisher Scientific™ Isotemp™ Refrigerator and Freezer Alarm Systems
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13-986-160 Freestanding alarm system
Each for $1,434.95
Check Availability
Add
to Cart
(0) in My Cart
13-986-100 Deluxe remote system
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Description
Freestanding Alarm System
  • Use with basic freezers, refrigerators, and one section on refrigerator/freezer combinations With key switch in ALARM position, system provides audio and visual alarms if chamber temperature reaches high or low alarm limit (adjustable, set by user) or if power is lost for more than 30 seconds
  • Housed in a painted metal cabinet with digital temperature display
  • Alarm silencing and test features
  • Can be used to trigger Standard or Deluxe Remote Alarm systems
Standard Remote System†
  • Use with refrigerators and freezers equipped with panel-mounted or freestanding alarm (Alarm or Recorder/Alarm models)
  • Can be located up to one-third mile from unit
  • Emits audible and visual signals in the event of power failure, temperature deviation, or if door is open for more than three minutes
  • Alarm sounds if line between unit and alarm is cut
Deluxe Remote System†
  • Use with refrigerators and freezers equipped with panel-mounted or freestanding alarm (Alarm or Recorder/Alarm models)
  • Can monitor up to four individual or four groups of refrigerators or freezers
  • User-programmable to call up to eight telephone or pager numbers when activated by power failure, temperature deviation, or if door is open for more than three minutes
  • Alarm sounds if line between unit and alarm is cut
  • Stores four outgoing messages
  • Automatic redial for busy or unanswered numbers
  • Works with any standard telephone system
  • Battery backup allows alarm to sound in the event of a power failure
Specifications & Ordering Information:
Freestanding and remote systems must be ordered when the refrigerator or freezer is ordered. Installation by owner.
† Requires an existing alarm system, with dry alarm contacts, to be installed on the refrigerator or freezer in order to be compatible.