Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
02-681-377
View Image Gallery (1) Image(s) are representative of the product Group
  • 02-681-377
Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Polypropylene Microtiter Tubes
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates
Also suitable for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagents. Choose plain tubes or tubes with frosted writing area.
  • Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
  • Nonsterile tubes are certified RNase- and DNase-free
  • Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
  • Frosted tubes feature writing area that wraps around tube collar
  • Capacity- 1.2mL
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Polypropylene Microtiter Tubes
  • 02-681-377
02-681-377
(10 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
1.2ml 96 Frostd Wr Tube 960/pk
02-681-382 N/AYou must be logged in and have a valid account number to view item availability   Case of 4800 for $304.00
(0) in My Cart
Plain tubes; Nonsterile; Tray pack
02-681-380 Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Pack of 960 for $70.30
Case of 5 PK for $300.00
(0) in My Cart
Frosted tubes; Nonsterile; Bagged
02-681-377 Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Pack of 1000 for $44.40
Case of 10 PK for $379.50
(0) in My Cart
Frosted tubes; Nonsterile; Robotic pack
02-681-387 Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Pack of 960 for $69.30
Case of 5 PK for $295.50
(0) in My Cart
Plain tubes; Nonsterile; Robotic pack
02-681-385 Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Pack of 960 for $69.30
Case of 5 PK for $295.50
(0) in My Cart
Frosted tubes; Sterile; Tray pack
02-681-383 Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Pack of 960 for $70.30
Case of 5 PK for $300.00
(0) in My Cart
Plain tubes; Sterile; Tray pack
02-681-381 Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Case of 4800 for $299.50
(0) in My Cart
Plain tubes; Sterile; Robotic pack
02-681-386 N/AYou must be logged in and have a valid account number to view item availability   Pack of 960 for $75.50
Case of 5 PK for $322.00
(0) in My Cart
Plain tubes; Nonsterile; Bagged
02-681-376 Available on GSA/VA Contract for Federal Government customers only.  N/AYou must be logged in and have a valid account number to view item availability   Pack of 1000 for $44.40
Case of 10 PK for $379.50
(0) in My Cart
1.2ml 96 Frostd Tubes 960/pk
02-681-388 N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Add Items to Cart
Description
  • Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
  • Nonsterile tubes are certified RNase- and DNase-free
  • Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
  • Frosted tubes feature writing area that wraps around tube collar
  • Capacity- 1.2mL
Specifications & Ordering Information:
Packed in bulk, or in convenient 8 x 12 compact tray packs or robotic racks.
Compact Tray Packs
  • Snug, smooth- fitting cover protects samples and enables stacking
  • With paper grid for easy indentification of contents
Robotic Racks
  • Specifically designed with 96-well footprint for robotic workstations
  • One-way lid design with alphanumeric grid enables easy sample identification
  • Condensation rings inside cover minimize cross-contamination
  • Tubes fit in straight, upright position
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.