Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
13-637-670
Available on GSA/VA Contract for Federal Government customers only. Fisher Scientific™ accumet™ Dot-Matrix Printer
Impact dot-matrix printer for accumet AR Series Benchtop Meters
Includes: 13-637-667 Serial AR Printer Cable
  • Permanently records results on 2.25 in. (5.7cm) paper with an ink ribbon
  • Prints up to 1.625 lines per second, 28.75 characters per line
  • Operates with large (12,500 lines) or small (3000 lines) roll paper
  • Print speed 38 lines per minute for 24-column
  • Printer communicates via standard RS-232-C interface (also includes RS485 port)
  • Baud rate is 19,200bps
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Fisher Scientific™ accumet™ Dot-Matrix Printer
  • 13-637-670
13-637-670
13-637-670
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13-637-668Available on GSA/VA Contract for Federal Government customers only.  Replacement ribbon
Each for $30.38
Check Availability
Add
to Cart
(0) in My Cart
13-637-669 Available on GSA/VA Contract for Federal Government customers only.  Printer paper
Each for $18.29
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Permanently records results on 2.25 in. (5.7cm) paper with an ink ribbon
  • Prints up to 1.625 lines per second, 28.75 characters per line
  • Operates with large (12,500 lines) or small (3000 lines) roll paper
  • Print speed 38 lines per minute for 24-column
  • Printer communicates via standard RS-232-C interface (also includes RS485 port)
  • Baud rate is 19,200bps
Specifications & Ordering Information:
Electrical Requirements: 120V 60Hz 20w (input to outlet); 9V 1.5A (output to printer). Stepdown converter included. Operating temperature, 5° to 40°C (41° to 104°F). Dimensions, 4.125L x 4.5W x 2 in.H (10.4 x 11.4 x 5cm).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.