Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. Sodium Lauryl Sulfate (Powder/NF/FCC), Fisher Chemical
Sodium Lauryl Sulfate
Powder
NF/FCC
Sodium Dodecyl Sulfate, SDS, Dodecylsulfuric Acid Sodium Salt, Duponol™ C Dry
C12H25NaO4S
CH3(CH2)10CH2OSO3Na
[151-21-3]
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Sodium Lauryl Sulfate (Powder/NF/FCC), Fisher Chemical
  • fisherchemical
(2 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
  • Special Offer
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S529-3 Available on GSA/VA Contract for Federal Government customers only.  Poly Pail; 3kg
MSDS
Each for $646.38
Each for $420.15 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
S529-500 Available on GSA/VA Contract for Federal Government customers only.  Glass Bottle; 500g
MSDS
Each for $152.05
Each for $98.83 Online Instant Savings (35%)
Case of 4 EA for $486.57
Case of 4 EA for $316.27 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
Go to Product Certificates
Product Specifications
Lead [Pb]5mg/kg max. (FCC)
AlkalinityPass Test (FCC/NF)
Total Alcohol59.0% min. (FCC/NF)
Heavy Metals [as Pb]0.002% max. (NF)
Sodium Chloride and Sodium Sulfate8.0% max. (FCC/NF)
Unsulfated Alcohols4.0% max. (FCC/NF)
Residual SolventsMeets Requirements (NF)
Flash Point: 100°C/212°F
Melting Point: 204° to 207°C
DANGER!
Emergency Overview
Flammable solid. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory reaction. Avoid contact with skin and eyes. Do not breathe dust. Use explosion-proof equipment. Use only non-sparking tools. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Remove from exposure, lie down. Move to fresh air.
NFPA
Health:2
Flammability:3
Instability:0