Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Thermo Scientific Spectronic Genesys 2 and Genesys 5 Spectrophotometer Softcard Software
Includes: All SoftCard™ software comes with memory data card, file transfer program and manual.
Application I SoftCard software
  • Performs standard curve, survey scan, three-point net absorbance, area-under-a-curve measurement, simple kinetics, and absorbance ratio functions
  • Application II SoftCard software
    • Performs same functions as Application I SoftCard software
    • Performs advanced kinetics, absorbance difference and multiple wavelength programs
    Biochemistry SoftCard software
    • Includes standard curve programs for determination of protein concentration by BCA and Bradford methods; one-step A260/A280 absorbance measurement and calculation program to determine DNA purity, DNA and protein concentrations; and chromatography column elution monitoring program
  • Performance Validation SoftCard software
    • Performs wavelength accuracy, noise, stray radiant energy, photometric linearity/accuracy, 0% transmission and output device diagnostics
    • Can be used with Thermo Spectronic standards set, 14-385-335, for ISO and GMP documentation
    Memory SoftCard data card
    • Store test set-up parameters, results and customized programs for up to 100 tests
    • Files stored in a DOS-compatible format, can be converted to ASCII or Lotus™ format for export to other software packages
    • 128kB, write protected supplied standard with all program software above and with GENESYS 2 spectrophotometer
    View Specifications
    Images for Thermo Scientific Spectronic Genesys 2 and Genesys 5 Spectrophotometer Softcard Software
    (1 item(s) in product Group)
    • Items
    • Specifications
    Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
    14-385-416
    Thermo Scientific
    No.:336090-20
    Memory Softcard
    N/A N/A N/A
    This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Application I SoftCard software
    • Performs standard curve, survey scan, three-point net absorbance, area-under-a-curve measurement, simple kinetics, and absorbance ratio functions
  • Application II SoftCard software
    • Performs same functions as Application I SoftCard software
    • Performs advanced kinetics, absorbance difference and multiple wavelength programs
    Biochemistry SoftCard software
    • Includes standard curve programs for determination of protein concentration by BCA and Bradford methods; one-step A260/A280 absorbance measurement and calculation program to determine DNA purity, DNA and protein concentrations; and chromatography column elution monitoring program
  • Performance Validation SoftCard software
    • Performs wavelength accuracy, noise, stray radiant energy, photometric linearity/accuracy, 0% transmission and output device diagnostics
    • Can be used with Thermo Spectronic standards set, 14-385-335, for ISO and GMP documentation
    Memory SoftCard data card
    • Store test set-up parameters, results and customized programs for up to 100 tests
    • Files stored in a DOS-compatible format, can be converted to ASCII or Lotus™ format for export to other software packages
    • 128kB, write protected supplied standard with all program software above and with GENESYS 2 spectrophotometer