Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
AC30161-5000
View Image Gallery (2) Image(s) are representative of the product Group
  • AC30161-5000
  • acros
a-Terpineol, 97+%, ACROS Organics™
a-Terpineol
97+%
p-Menth-1-en-8-ol
C10H18O
F.W.: 154.25
[98-55-5, 97.00-100.00%]
View Specifications
Images for a-Terpineol, 97+%, ACROS Organics™
  • AC30161-5000
  • acros
AC30161-5000
AC30161-5000
(3 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
  • Special Offer
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
AC301610250
ACROS Organics
No.:301610250
25g; Glass bottle
MSDS
Each for $49.00
Check Availability
Add
to Cart
(0) in My Cart
AC301615000
ACROS Organics
No.:301615000
500g; Glass bottle
MSDS
Each for $101.40
Check Availability
Add
to Cart
(0) in My Cart
AC30161-0025
ACROS Organics
No.:301610025
2.5kg
MSDS
Each for $115.24
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Product Specifications
Assay97.0% min.
Physical FormClear colorless liquid after melting
Specific Gravity(20°C) 0.934 to 0.940
Infrared SpectrumAuthentic
Refractive Index1.4800 to 1.4830 (20°C, 589nm)
Boiling Point: 214.0°C to 224.0°C
Density: 0.9370g/mL
Flash Point: 90°C
Melting Point: 31.0°C to 34.0°C
n: 1.4800 to 1.4830
Solubility in water: insoluble. Other solubilities: soluble in organic solvents , diethyl ether,, ethanol, benzene, propylene glycol
Sp. gr.: 0.9370
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.