Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
22-024-850
Abaxis Piccolo™ xpress Clinical Chemistry Analyzer
On-the-spot chemistry results in minutes
  • Fully automated system with walk-away operation
  • Uses a small sample size (0.1cc of whole blood, serum, or plasma)
  • No sample pre-treatment is needed
  • Results available in approximately 12 minutes
  • Built-in intelligent quality control meets CLIA and NCCLS requirements for daily control testing
  • Delivers up to 24 individual tests including CLIA-waived lipids, liver enzymes, and glucose monitoring
  • Results are provided in a “hard copy” self-adhesive report or can be quickly transferred to a printer, computer, or data management system
  • Small footprint measures 32.4cm (12.75 in.) x 15.2cm (6 in.) x 20.3cm (8 in.)
  • Portable and lightweight; with carrying handle only weighs 11.2 lb.
  • Bar coded self-contained reagent discs are self-calibrating
  • System stores up to 5000 patient and control results on-board
  • Easy-to-use color touchscreen
View Specifications
Images for Abaxis Piccolo™ xpress Clinical Chemistry Analyzer
  • 22-024-850
22-024-850
22-024-850
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-024-850
Abaxis
No.:11000000
Cholesterol Analyzer
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
  • Fully automated system with walk-away operation
  • Uses a small sample size (0.1cc of whole blood, serum, or plasma)
  • No sample pre-treatment is needed
  • Results available in approximately 12 minutes
  • Built-in intelligent quality control meets CLIA and NCCLS requirements for daily control testing
  • Delivers up to 24 individual tests including CLIA-waived lipids, liver enzymes, and glucose monitoring
  • Results are provided in a “hard copy” self-adhesive report or can be quickly transferred to a printer, computer, or data management system
  • Small footprint measures 32.4cm (12.75 in.) x 15.2cm (6 in.) x 20.3cm (8 in.)
  • Portable and lightweight; with carrying handle only weighs 11.2 lb.
  • Bar coded self-contained reagent discs are self-calibrating
  • System stores up to 5000 patient and control results on-board
  • Easy-to-use color touchscreen