Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (6) Image(s) are representative of the product Group
  • 22037738
  • 22272483
  • 22269973
  • 22383380
  • 22037734
Available on GSA/VA Contract for Federal Government customers only. Medical Packaging Pap-Pak™ Pap Smear Collection System
Each kit contains all the necessary materials for specimen collection, smear preparation, fixation and transportation to the laboratory
  • Contains the 8 in. (20cm)CytoSoft™ cytology brush that enhances retrieval of endocervical cells and reduces the possibility of false negative smears
  • Pap-Pak standardizes specimen collection, provides illustrated instructions, complies with federal quality assurance regulations and reduces costs by eliminating waste and misuse of cytology supplies
  • Each kit is supplied with the option of one foil pouch fixative per kit or twenty 1 oz. (30mL) pump spray fixative bottles per case
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Medical Packaging Pap-Pak™ Pap Smear Collection System
  • 22037738
  • 22272483
  • 22269973
  • 22383380
  • 22037734
(6 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-037-734
MPC
No.:S-100P-1
Pappak 1sld W/1oz Pmp Fx 25/pk
Accessories
Pack of 25 for $19.89
Pack of 25 for $12.93 Online Instant Savings (35%)
Case of 20 PK for $331.42
Case of 20 PK for $215.42 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
22-037-738 Available on GSA/VA Contract for Federal Government customers only. 
MPC
No.:C-100P1
Without Mailing Bracket; 1 slide; Pump Spray
Accessories
Pack of 25 for $33.05
Pack of 25 for $21.48 Online Instant Savings (35%)
Case of 20 PK for $661.16
Case of 20 PK for $429.75 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
22-037-740 Available on GSA/VA Contract for Federal Government customers only. 
MPC
No.:C-500P-1
With Mailing Bracket; 1 slide; Pump Spray
Accessories
Pack of 25 for $38.14
Pack of 25 for $24.79 Online Instant Savings (35%)
Case of 20 PK for $762.84
Case of 20 PK for $495.85 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
22-269-973 Available on GSA/VA Contract for Federal Government customers only. 
MPC
No.:C100
Without mailing bracket; 1 slide; Foil pouch
Accessories
Pack of 25 for $32.99
Pack of 25 for $21.44 Online Instant Savings (35%)
Case of 20 PK for $657.76
Case of 20 PK for $427.54 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
22-272-483 Available on GSA/VA Contract for Federal Government customers only. 
MPC
No.:C-500
With Mailing Bracket; 1 slide; Foil Pouch
Pack of 25 for $36.20
Pack of 25 for $23.53 Online Instant Savings (35%)
Case of 20 PK for $724.08
Case of 20 PK for $470.65 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
22-383-380
MPC
No.:S-100
Pappak Std Kit 1slid Vce 25/pk
Pack of 25 for $22.05
Pack of 25 for $14.33 Online Instant Savings (35%)
Case of 20 PK for $377.10
Case of 20 PK for $245.12 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Contains the 8 in. (20cm)CytoSoft™ cytology brush that enhances retrieval of endocervical cells and reduces the possibility of false negative smears
  • Pap-Pak standardizes specimen collection, provides illustrated instructions, complies with federal quality assurance regulations and reduces costs by eliminating waste and misuse of cytology supplies
  • Each kit is supplied with the option of one foil pouch fixative per kit or twenty 1 oz. (30mL) pump spray fixative bottles per case
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.