Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Tubes, Petters and Caps
Available on GSA/VA Contract for Federal Government customers only. KOVA International KOVA™ Tubes, Petters and Caps
Help guard against sample loss during transporting, centrifuging, and decanting
  • Super Tubes are marked in 1mL graduations, made of K-Resin™ and have a 12mL capacity Economy Tubes are made of polystyrene and have a 12mL capacity
  • Decanting Petter is a multipurpose plastic transfer pipet for contamination-free decanting
  • Locking snugly into a KOVA Tube during decanting, the Petter retains precisely 1mL of urine sediment and gently re-suspends and transfers it onto the microscope slide for examination Tube Caps fit both Super and Economy Tubes described above, provide secure closures during transport and storage and prevent aerosoling during centrifugation of potentially infectious specimens
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. KOVA International KOVA™ Tubes, Petters and Caps
  • Tubes, Petters and Caps
Tubes, Petters and Caps
Tubes, Petters and Caps
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-741-165 Available on GSA/VA Contract for Federal Government customers only. 
KOVA International
No.:87137
Super Tubes
Accessories
VT87135 Available on GSA/VA Contract for Federal Government customers only. 
KOVA International
No.:87135
Decanting Petter
VT87138 Available on GSA/VA Contract for Federal Government customers only. 
KOVA International
No.:87138
Economy Tubes
Accessories
VT87139 Available on GSA/VA Contract for Federal Government customers only. 
KOVA International
No.:87139
Tube Caps
Description
  • Super Tubes are marked in 1mL graduations, made of K-Resin™ and have a 12mL capacity Economy Tubes are made of polystyrene and have a 12mL capacity
  • Decanting Petter is a multipurpose plastic transfer pipet for contamination-free decanting
  • Locking snugly into a KOVA Tube during decanting, the Petter retains precisely 1mL of urine sediment and gently re-suspends and transfers it onto the microscope slide for examination Tube Caps fit both Super and Economy Tubes described above, provide secure closures during transport and storage and prevent aerosoling during centrifugation of potentially infectious specimens
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.