Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S49080
Products for Science Education Clay Adams™ Compact II Centrifuge
Safety shield protects operators from accidents.
Includes:
  • Stainless-steel shields with cushions
  • 12 adapters
  • 6 ft. (1.8m) heavy-duty cord
  • Six-place angle head centrifuge with safety shield
  • Runs at fixed 3200rpm; 3060 to 3400rpm range
  • 30-minute timer with hold
  • Capacity: 90mL (6 x 15mL)
  • Adapters allow 5mL taper-bottom or 13 x 100mm or 16 x 100mm culture tubes
  • Dimensions: 7.5 dia. x 10 in.H (19.0 x 25.4cm)
  • For 120V 60Hz, 1.5A
View Specifications
Images for Products for Science Education Clay Adams™ Compact II Centrifuge
  • S49080
S49080
S49080
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S49080
BD
No.:420225/EMD
Compact Centrifuge; 6 places
Accessories
Each for $1,505.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Six-place angle head centrifuge with safety shield
  • Runs at fixed 3200rpm; 3060 to 3400rpm range
  • 30-minute timer with hold
  • Capacity: 90mL (6 x 15mL)
  • Adapters allow 5mL taper-bottom or 13 x 100mm or 16 x 100mm culture tubes
  • Dimensions: 7.5 dia. x 10 in.H (19.0 x 25.4cm)
  • For 120V 60Hz, 1.5A