Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S11509E
Products for Science Education Edvotek™ EVT 300 Dual Power Supply
Power for one or two electrophoresis units.
  • Designed for safety
  • Delivers direct current, constant voltage at settings of 75 and 150V
  • Maximum current is 250mA
View Specifications
Images for Products for Science Education Edvotek™ EVT 300 Dual Power Supply
  • S11509E
S11509E
S11509E
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S11509E
Edvotek Inc.
No.:509
EVT 300
Description
  • Designed for safety
  • Delivers direct current, constant voltage at settings of 75 and 150V
  • Maximum current is 250mA