Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
22-143-975
Available on GSA/VA Contract for Federal Government customers only. Citrisolv Clearing Agent, Fisherbrand
Fisherbrand™ CitriSolv™ Clearing Agent
  • Xylene substitute for use in histology, cytology and parasitology
  • Pleasant-smelling, limonene-based solvent derived from citrus
  • Soluble with absolute alcohol, paraffin embedding media, and mounting media
  • Causes minimal tissue shrinkage and actually enhances some stains
  • Dissolves paraffin waxes, glues, and adhesives; dries slowly with no residue
  • Can safely be used in most histology, cytology, hematology, and microbiology procedures that require xylene
  • Biodegradable
[5989-27-5, 97.00-98.00%], [25013-16-5, 0.20-0.24%]
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Images for Available on GSA/VA Contract for Federal Government customers only. Citrisolv Clearing Agent, Fisherbrand
  • 22-143-975
22-143-975
22-143-975
(1 item(s) in product Group)
  • Items
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-143-975 Available on GSA/VA Contract for Federal Government customers only.  Clearing agent; 1 gal.
Each for $252.50
Case of 4 EA for $512.50