Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (2) Image(s) are representative of the product Group
  • fisherchemical
Methanol (HPLC), Fisher Chemical
Methanol
HPLC Grade, Mobile phase for HPLC applications.
Assay: 99.9% min by GC
HPLC mobile phase for analytical and preparative separations.
High UV transmittance and low evaporation residue.
Suitable for gradient and isocratic conditions.
Filtered through a 0.2µm filter to remove particulates.
This product also meets ACS specifications.
MeOH, Methyl Alcohol
CH4O
CH3OH
F.W.: 32.04
[67-56-1]
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
View Specifications
Images for Methanol (HPLC), Fisher Chemical
  • fisherchemical
(5 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
A452-SK4 Available on GSA/VA Contract for Federal Government customers only.  Glass Bottle, Safecote; 4L
MSDS
Each for $125.18
Each for $75.11 Online Instant Savings (40%)
Case of 4 EA for $400.57
Case of 4 EA for $240.34 Online Instant Savings (40%)
Check Availability
Add
to Cart
(0) in My Cart
A452SK-1 Available on GSA/VA Contract for Federal Government customers only.  Glass Bottle, Safecote; 1L
MSDS
Each for $92.96
Each for $55.78 Online Instant Savings (40%)
Case of 6 EA for $418.33
Case of 6 EA for $251.00 Online Instant Savings (40%)
Check Availability
Add
to Cart
(0) in My Cart
A452N1-19 Available on GSA/VA Contract for Federal Government customers only.  NOWPAK I; 19L
MSDS
Each for $617.05
Check Availability
Add
to Cart
(0) in My Cart
A452-4 Available on GSA/VA Contract for Federal Government customers only.  Glass Bottle; 4L
MSDS
Each for $110.12
Each for $66.07 Online Instant Savings (40%)
Case of 4 EA for $352.38
Case of 4 EA for $211.43 Online Instant Savings (40%)
Check Availability
Add
to Cart
(0) in My Cart
A452-1 Available on GSA/VA Contract for Federal Government customers only.  Glass Bottle; 1L
MSDS
Each for $76.75
Each for $46.05 Online Instant Savings (40%)
Case of 6 EA for $345.36
Case of 6 EA for $207.22 Online Instant Savings (40%)
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Go to Product Certificates
0.2-micron filtered.
Product Specifications
Assay99.9% min.
Color5 APHA max.
Titratable Acid0.0003mEq/g max.
Absorbance at 205nm1.00 AU max.
Absorbance at 220nm0.30 AU max.
Absorbance at 230nm0.15 AU max.
Absorbance at 254nm0.025 AU max.
Carbonyl Compounds0.001% max.
IdentificationPass Test
Refractive Index1.3260 to 1.3300 (at 25°C)
Residue after Evaporation3ppm max.
Titratable Base0.0002mEq/g max.
Solubility in WaterPass Test
Substances Darkened by H2SO4Pass Test
Substances Reducing PermanganatePass Test
Water0.1% max.
Fluorescence BackgroundPass Test
Additional InformationVapor Pressure: 128hPa at 20°C
Boiling Point: 64.7°C (148.5°F)
Density: 1.11 (Air = 1.0)
Flash Point: 12°C/53.6°F
Melting Point: -98°C (-144.4°F)
Miscible with water
Sp. gr.: 0.791
0.55cp at 20°C
 solvent  
1230
DANGER!
Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and eyes. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.
NFPA
Health:1
Flammability:3
Instability:0