Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
09-898-12A
Fisherbrand™ Low-Nitrogen Weighing Paper
For analytical balances
  • Sufficient body to support sample
  • Smooth textured to avoid trapping material
  • Does not absorb moisture
View Specifications
Images for Fisherbrand™ Low-Nitrogen Weighing Paper
  • 09-898-12A
09-898-12A
09-898-12A
(3 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
09-898-12A 3 x 3 in. (76 x 76mm)
Pack of 500 for $37.30
Case of 12 PK for $370.26
Check Availability
Add
to Cart
(0) in My Cart
09-898-12B 4 x 4 in. (100 x 100mm)
Pack of 500 for $37.12
Case of 12 PK for $380.92
Check Availability
Add
to Cart
(0) in My Cart
09-898-12C 6 x 6in. (152 x 152mm)
Pack of 500 for $37.87
Case of 12 PK for $388.40
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Sufficient body to support sample
  • Smooth textured to avoid trapping material
  • Does not absorb moisture