Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical
Platinum Cobalt Color Standard Solution
APHA No. 500 Color Standard for water and clear liquids. Meets ASTM D1209-00.
Conforms to APHA standard.
[7647-01-0], [7791-13-1], [16921-30-5], [7732-18-5]
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
View Specifications
Images for Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical
  • fisherchemical
(2 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
SP120-1 Glass Bottle; 1L
MSDS
Each for $685.40
Case of 6 EA for $3,084.31
Check Availability
Add
to Cart
(0) in My Cart
SP120-500 Glass Bottle; 500mL
MSDS
Each for $394.54
Case of 6 EA for $1,775.45
Check Availability
Add
to Cart
(0) in My Cart
Go to Product Certificates
Product Specifications
Absorbance at 430nm0.110 to 0.120 AU
Absorbance at 455nm0.130 to 0.145 AU
Absorbance at 480nm0.105 to 0.120 AU
Absorbance at 510nm0.055 to 0.065 AU
IdentificationPass Test
Density: 1g/mL
Melting Point: 0°C
1789