Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Science Project Kits
View Image Gallery (4) Image(s) are representative of the product Group
  • Science Project Kits
  • S66216
  • S66218
  • S66219
Products for Science Education Science Project Kits
Excellent ideas for science fair projects.
Help students to develop ideas for science fair projects using these kits. The Science Project Kits guide students through the entire project, from choosing a subject, doing the research, collecting the data, and finally displaying the results. TesTabs™ reagents, test tubes, and project ideas are included.
View Specifications
Images for Products for Science Education Science Project Kits
  • Science Project Kits
  • S66216
  • S66218
  • S66219
Science Project Kits
Science Project Kits
(3 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S66216
Lamotte Chemical Products Company
No.:3-5873-6
pH Science Project Kit
Each for $2.75
Case of 6 EA for $16.50
Check Availability
Add
to Cart
(0) in My Cart
S66218
Lamotte Chemical Products Company
No.:3-5875-6
Bacteria Science Project Kit
Each for $2.75
Case of 6 EA for $16.50
Check Availability
Add
to Cart
(0) in My Cart
S66219
Lamotte Chemical Products Company
No.:3-5876-6
Chlorine Science Project Kit
Each for $2.75
Case of 6 EA for $16.50
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
Help students to develop ideas for science fair projects using these kits. The Science Project Kits guide students through the entire project, from choosing a subject, doing the research, collecting the data, and finally displaying the results. TesTabs™ reagents, test tubes, and project ideas are included.
pH Science Project Kit. Topics include acid rain, corrosion, and acids and bases. Create an experiment to test household items and make a pH scale. 20 tests.
Bacteria Science Project Kit. Explore how water is tested to make sure it is safe for drinking and swimming. Test water samples for the presence of coliform bacteria. Three tests.
Chlorine Science Project Kit. Find out how water is treated to make it cleaner for drinking and swimming. Use TesTabs reagents to measure water samples for safe levels of sanitizers in swimming pools and other water samples. 20 tests.
Copper/Iron Science Project Kit. Find out why corrosion causes damage to water systems and metal surfaces. Demonstrate corrosion and measure the amount of copper and iron in water samples. Ten tests for copper and ten tests for iron.