Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S65959
Available on GSA/VA Contract for Federal Government customers only. Products for Science Education Kontes™ Kem-Kit™ Semi-Macro 19/22 Organic Chemistry Kit
Compact, 14-piece kit for organic chemistry studies
  • Selection of basic small-scale glassware with 19/22 joints
  • Performs extraction, reflux, reflux with addition, fractionation, distillation (steam, atmospheric, and vacuum), and other organic chemistry applications
  • Supplied in a heavy-duty, double-wall polyethylene storage case with handle and hinged lid
  • Case dimensions: 13.5L x 11W x 5.5 in.H (34 x 28 x 14cm)
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Products for Science Education Kontes™ Kem-Kit™ Semi-Macro 19/22 Organic Chemistry Kit
  • S65959
S65959
S65959
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S65959 Available on GSA/VA Contract for Federal Government customers only. 
Kimble Chase Kontes
No.:269800-0000EMD
Kem Kit Semi-macro 19/22
Each for $630.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Selection of basic small-scale glassware with 19/22 joints
  • Performs extraction, reflux, reflux with addition, fractionation, distillation (steam, atmospheric, and vacuum), and other organic chemistry applications
  • Supplied in a heavy-duty, double-wall polyethylene storage case with handle and hinged lid
  • Case dimensions: 13.5L x 11W x 5.5 in.H (34 x 28 x 14cm)
Specifications & Ordering Information:
  • Comprehensive instruction manual
  • Storage case
DescriptionKontes No.
Bleed Adapter273410-0000
Claisen Adapter273760-0000
Connecting Adapter275060-0000
Outlet Adapter275410-1922
Vacuum Adapter276410-0000
Condenser, West282460-0200
Distillation Column286810-0200
Flask, 25mL294010-0025
Flask, 50mL294010-0050
Flask, 100mL294010-0100
Flask, 250mL294010-0250
Flask, 500mL294010-0500
Funnel, 125mL, PTFE298290-0125
Stopper850500-1922
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.