Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Revco Pharmacy Refrigerators
View Image Gallery (2) Image(s) are representative of the product Group
  • Revco Pharmacy Refrigerators
  • 13-991-202
Thermo Scientific™ Revco™ Pharmacy Refrigerators
Designed for accurate storage and easy retrieval of pharmacy products
Microprocessor Control
  • Adjustable temperature control from 1° to 8°C, factory preset at 4°C
  • Icon-based controls and displays include graphic references for universal operation
  • Setpoint security with key-operated, triple-position master switch controls power to all systems
  • Audible/visual warnings for overtemperature, undertemperature and power failure, with visual status reports on critical functions
  • Graphic thermometer confirms proper operation: steady green for normal, flashing top bar for overtemperature, flashing bulb for undertemperature
  • Advanced defrost sensor manages the defrost cycle, minimizing frost buildup
  • Enlarged digital display with resolution to within 0.1°C
  • Alarm silence, ringback and automatic reset functions
  • Push-button alarm test verifies sensor continuity
  • Battery backup assures control panel function during power failure
  • Remote alarm contacts
Refrigeration System
  • Heavy-duty, industrial-grade, hermetically sealed compressors
  • Automatic defrost maintains optimum cooling capacity
  • Automatic condensate removal; no drain lines required
View Specifications
Images for Thermo Scientific™ Revco™ Pharmacy Refrigerators
  • Revco Pharmacy Refrigerators
  • 13-991-202
Revco Pharmacy Refrigerators
Revco Pharmacy Refrigerators
(7 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13-991-202 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RPR1204A
11.5 cu. ft., 115V 60Hz
Each for $5,800.00
Check Availability
Add
to Cart
(0) in My Cart
13-991-200 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RPR2304A
23.3 cu. ft., 115V 60hz
Each for $7,841.00
Check Availability
Add
to Cart
(0) in My Cart
13-991-201 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RPR2304D
23.3 cu. ft., 208/230V 60Hz
Each for $7,841.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
13-991-133
Thermo Scientific
No.:RPR3004A
29.2 cu. ft.; 115V 60Hz
Each for $9,011.00
Check Availability
Add
to Cart
(0) in My Cart
13-991-134 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RPR3004D
29.2 cu. ft.; 208/230V 60Hz
Each for $9,011.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
13-991-204 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RPR5004A
51.1 cu.ft; 115V 60Hz
Each for $11,671.00
Check Availability
Add
to Cart
(0) in My Cart
13-991-205 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RPR5004D
51.1 cu.ft; 208/230V 60Hz
Each for $11,671.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add Items to Cart
Description
Microprocessor Control
  • Adjustable temperature control from 1° to 8°C, factory preset at 4°C
  • Icon-based controls and displays include graphic references for universal operation
  • Setpoint security with key-operated, triple-position master switch controls power to all systems
  • Audible/visual warnings for overtemperature, undertemperature and power failure, with visual status reports on critical functions
  • Graphic thermometer confirms proper operation: steady green for normal, flashing top bar for overtemperature, flashing bulb for undertemperature
  • Advanced defrost sensor manages the defrost cycle, minimizing frost buildup
  • Enlarged digital display with resolution to within 0.1°C
  • Alarm silence, ringback and automatic reset functions
  • Push-button alarm test verifies sensor continuity
  • Battery backup assures control panel function during power failure
  • Remote alarm contacts
Refrigeration System
  • Heavy-duty, industrial-grade, hermetically sealed compressors
  • Automatic defrost maintains optimum cooling capacity
  • Automatic condensate removal; no drain lines required
Construction
  • Adjustable full-extension stainless-steel drawers
  • Full-view glass door(s): double-pane construction eliminates condensation
  • Spring-loaded door closures enable 90° stay-open
  • High-density, CFC-free urethane foam insulation protects cabinet from ambient temperature fluctuations
  • Positive, forced-air circulation maintains temperature uniformity at all shelf levels
  • Directional airflow assures quick recovery after door openings
  • Interior cabinet lights with door-activated ON/OFF switch, plus independent switch on control panel
  • Doors self-close against closed-cell peripheral gasket
  • Door-locking provision
  • Adjustable leveling feet (optional casters available)
  • Smooth, scratch-resistant painted interior and exterior surfaces
Specifications
Temperature Range1° to 8°C
Temperature Resolution0.1°C
ControlMicroprocessor
DisplayDigital
DefrostAutomatic
RefrigerantCFC-free
CabinetSmooth, scratch-resistant painted
ChamberSmooth, scratch-resistant painted
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.