Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Immu-Mount
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific Shandon™ Immu-Mount™
  • Non-permanent mountant
  • Economical, organic, overnight-drying liquid coverslip
  • Aqueous base for immunostaining and other techniques
  • Non-fluorescing medium with a low viscosity
  • Resists fading and polymerizes quickly for easier handling of finished slides
  • Refractive index: 1.495 ±0.002 (liquid); 1.586 ± 0.002 (dry)
  • Viscosity: Low (120-160 cps at 24°C)
  • pH: 8.0 from 8.8 at 21° through 24°C
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific Shandon™ Immu-Mount™
  • Immu-Mount
Immu-Mount
Immu-Mount
(3 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
99-904-02 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:9990402
20mL x 1
MSDS
Each for $22.75
Each for $19.34 Online Instant Savings (15%)
Check Availability
Add
to Cart
(0) in My Cart
99-904-12 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:9990412
20mL x 6
MSDS
Case of 6 for $110.98
Case of 6 for $94.33 Online Instant Savings (15%)
Check Availability
Add
to Cart
(0) in My Cart
99-904-14 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:9990414
1L
MSDS
Each for $361.37
Each for $307.16 Online Instant Savings (15%)
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Non-permanent mountant
  • Economical, organic, overnight-drying liquid coverslip
  • Aqueous base for immunostaining and other techniques
  • Non-fluorescing medium with a low viscosity
  • Resists fading and polymerizes quickly for easier handling of finished slides
  • Refractive index: 1.495 ±0.002 (liquid); 1.586 ± 0.002 (dry)
  • Viscosity: Low (120-160 cps at 24°C)
  • pH: 8.0 from 8.8 at 21° through 24°C
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.