Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S440971
Products for Science Education Geiger Counter
Large four-digit readout shows counts per minute or continuous counting.
  • Student-ready, compact and rugged four-digit Geiger counter
  • Suitable for the Geiger stand with absorbers (S440974)
  • Indicator lamp and audible chirp signal show tube counts
  • Internal power supply will operate the 900VDC Geiger tube (S440972) or the smaller 450VDC tube (S440973)
  • Front panel has a power switch and a switch to select counts per minute or continuous counting
  • Push-button for manual resetting is provided
  • Rear panel has connectors for the GM tube and a mute switch for the chirp signal
View Specifications
Images for Products for Science Education Geiger Counter
  • S440971
S440971
S440971
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S440971
Science Source
No.:EN-15
Geiger Counter
Each for $651.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Student-ready, compact and rugged four-digit Geiger counter
  • Suitable for the Geiger stand with absorbers (S440974)
  • Indicator lamp and audible chirp signal show tube counts
  • Internal power supply will operate the 900VDC Geiger tube (S440972) or the smaller 450VDC tube (S440973)
  • Front panel has a power switch and a switch to select counts per minute or continuous counting
  • Push-button for manual resetting is provided
  • Rear panel has connectors for the GM tube and a mute switch for the chirp signal
Specifications & Ordering Information:
Dimensions: 3.5 x 6.5 x 5.5 in. (8.9 x 16.5 x 14cm).