Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S40641P
Products for Science Education Fisher Scientific™ Measuring Ruler
Useful conversion tables make this a valuable laboratory tool.
  • Inexpensive, blue vinyl
  • Graduated from 0 to 6 in. and 0 to 150mm
  • Temperature conversion scale from -20° to +100°C, and -10° to +212°F
  • Scale for quickly determining cork and rubber stopper sizes
View Specifications
Images for Products for Science Education Fisher Scientific™ Measuring Ruler
  • S40641P
S40641P
S40641P
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
S40641 Blue Vinyl 100/pk
Pack of 100 for $49.00
Check Availability
Add
to Cart
(0) in My Cart
S40641P Blue vinly; Plastic; 6 in.(15cm) long
Pack of 10 for $7.70
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Inexpensive, blue vinyl
  • Graduated from 0 to 6 in. and 0 to 150mm
  • Temperature conversion scale from -20° to +100°C, and -10° to +212°F
  • Scale for quickly determining cork and rubber stopper sizes