Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Lonza BioWhittaker™ RPMI 1640 with L-Glutamine
Manufactured using formulations recommended by the Tissue Culture Association and Water for Injection (WFI) quality water. Sterile-filtered through two consecutive 0.2µm pharmaceutical-grade filters.
View Specifications
Images for Lonza BioWhittaker™ RPMI 1640 with L-Glutamine
(11 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
BW12-115Q
Lonza
No.:12115Q
w/ L-Glutamine and 25mM HEPES Buffer; Quantity: 1L
Each for $52.41
Check Availability
Add
to Cart
(0) in My Cart
BW12702Q12
Lonza
No.:12702Q12
w/ L-Glutamine; Quantity: 12 x 1L
Pack of 12 for $358.32
Check Availability
Add
to Cart
(0) in My Cart
BW12-115F
Lonza
No.:12115F
w/ L-Glutamine and 25mM HEPES Buffer; Quantity: 500mL
Each for $33.57
Check Availability
Add
to Cart
(0) in My Cart
BW09-774E
Lonza
No.:09-774E
w/ L-Glutamine, 25mM HEPES Buffer, 100 units Penicillin/50mg Streptomycin per mL; 100mL
Each for $16.87
Check Availability
Add
to Cart
(0) in My Cart
BW12-702Q
Lonza
No.:12702Q
w/ L-Glutamine; Quantity: 1L
Each for $33.44
Check Availability
Add
to Cart
(0) in My Cart
BW12702F12
Lonza
No.:12-702F/12
w/ L-Glutamine; Quantity: 12 x 500mL
Pack of 12 for $218.59
Check Availability
Add
to Cart
(0) in My Cart
BW12-702F
Lonza
No.:12702F
w/ L-Glutamine; Quantity: 500mL
Each for $21.54
Check Availability
Add
to Cart
(0) in My Cart
BW12115Q12
Lonza
No.:12-115Q/12
w/ L-Glutamine and 25mM HEPES Buffer; Quantity:12 x 1L
Pack of 12 for $513.91
Check Availability
Add
to Cart
(0) in My Cart
BW12115F12
Lonza
No.:12115F12
w/ L-Glutamine and 25mM HEPES Buffer; 12 x 500mL
Pack of 12 for $318.27
Check Availability
Add
to Cart
(0) in My Cart
BW09-774F
Lonza
No.:09-774F
w/ L-Glutamine, 25mM HEPES Buffer, 100 units Penicillin/50mg Streptomycin per mL; 500mL
Each for $36.19
Check Availability
Add
to Cart
(0) in My Cart
BW04-525F
Lonza
No.:04-525F
w/L-Glutamine, w/o Sodium Bicarbonate and MOPS; 500mL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Add Items to Cart
Manufactured using formulations recommended by the Tissue Culture Association and Water for Injection (WFI) quality water. Sterile-filtered through two consecutive 0.2µm pharmaceutical-grade filters.
Recommended Storage: Store as detailed on the label.