Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S63038
Products for Science Education Heidolph™ Tuttnauer™ Autoclaves
Slow and fast exhaust in one unit.
Heidolph Tuttnuaer autoclaves provide fast, safe, dependable and convenient means of sterilizing liquids, media, instruments, glassware, clothing and waste. Tuttnauer provides a line with a wide range of models from our economic manually controlled analog models to our electronically controlled models with a built-in printer for GLP-compliant documentation.
Standard Operating Features
  • Self-contained design eliminates external plumbing and hardwiring for easy installation
  • Simple Operation
  • ASME- stamped, 316L stainless steel chamber ensures long-lasting reliability, durability and high corrosions resistance
  • Fast/slow exhaust; fast exhaust allows quick cycle time for solids and slow exhaust prevents media and liquid boil-over
  • Closed-door drying system maintains sterility
  • Drying cycle removes residual moisture after sterilization
  • Continuous pressure purge eliminates air pockets and ensures through, even sterilization
  • Large-capacity reservoir provides an ample water supply to avoid frequent fill-ups; the water drain in conveniently located on top to allow for quick and easy cleaning of the reservoir
  • Automatic shut-off feature saves energy
  • One-year warranty
  • Standard Safety Features
    • Heat-insulated, double-locking door protects against accidental burns; the door cannot be opened while the chamber is under pressure
    • Automatic low water level shut-off prevents damage to the heater elements if there is insufficient water for the sterilization cycle
    • Over-temperature cut-off and over-pressure valve ensure safe operation
    • UL- and CSA- approved
    Economical Analog Sterilizers
    • These economical, manually operated models feature analog controls and gauges for simple operation
    • 60-minute cycle timer
    • Temperature selection from 1000°C to 1340°C
    • Combination pressure and temperature gauge with peak indicator
    View Specifications
    Images for Products for Science Education Heidolph™ Tuttnauer™ Autoclaves
    • S63038
    S63038
    S63038
    (4 item(s) in product Group)
    • Items
    • Specifications
    Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
    S63039
    Heidolph North America
    No.:023210100EMD
    Analog Control, 9 in. I.D.; 120V
    Accessories
    Each for $7,321.00
    Check Availability
    Add
    to Cart
    (0) in My Cart
    S63100A
    Heidolph North America
    No.:023210401/EMD
    Electronic Controls, 10 in. I.D., four shelves; 120V
    Each for $8,411.00
    Check Availability
    Add
    to Cart
    (0) in My Cart
    S63038
    Heidolph North America
    No.:23210002 EMD
    Analog Control, 7 in. I.D., three shelves; 120V
    Accessories
    N/A N/A N/A
    This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
    View substitute products »
    S63040
    Heidolph North America
    No.:23210151 EMD
    Analog Control, 9 in. I.D.; 220V
    Accessories
    N/A N/A N/A
    This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
    View substitute products »
    Add Items to Cart
    Description
    Standard Operating Features
    • Self-contained design eliminates external plumbing and hardwiring for easy installation
    • Simple Operation
    • ASME- stamped, 316L stainless steel chamber ensures long-lasting reliability, durability and high corrosions resistance
    • Fast/slow exhaust; fast exhaust allows quick cycle time for solids and slow exhaust prevents media and liquid boil-over
    • Closed-door drying system maintains sterility
    • Drying cycle removes residual moisture after sterilization
    • Continuous pressure purge eliminates air pockets and ensures through, even sterilization
    • Large-capacity reservoir provides an ample water supply to avoid frequent fill-ups; the water drain in conveniently located on top to allow for quick and easy cleaning of the reservoir
    • Automatic shut-off feature saves energy
    • One-year warranty
  • Standard Safety Features
    • Heat-insulated, double-locking door protects against accidental burns; the door cannot be opened while the chamber is under pressure
    • Automatic low water level shut-off prevents damage to the heater elements if there is insufficient water for the sterilization cycle
    • Over-temperature cut-off and over-pressure valve ensure safe operation
    • UL- and CSA- approved
    Economical Analog Sterilizers
    • These economical, manually operated models feature analog controls and gauges for simple operation
    • 60-minute cycle timer
    • Temperature selection from 1000°C to 1340°C
    • Combination pressure and temperature gauge with peak indicator
    Warranty: Two years
    Autoclave Holds125mL Erlenmeyer Flasks500mL Erlenmeyer Flasks1L, short neck2L, short neck500mL Media Bottle
    S630385----------------
    S6303912----------------
    S6304012----------------
    S63100A1243----9