Log into Your Account
Don't have a profile? Register
Forgot Password?
Search
Customer Service:  800-766-7000Web Support:  877-885-2081All Other Contact Info
 Shopping Cart View Cart 
No items in cart
For security reasons you will be automatically logged out after 30 minutes of inactivity.
Products for Science Education Ealing Air Table
More accurate and durable than conventional air tables!
The extremely flat surface (kept below 0.0254cm) and resistance to warping improve your results and add years to the life to your air table. Applications include the study of Newton's laws of motion, conservation of momentum, collisions, and many more!
See details
Includes: Low K springs, six pucks, Velcro puck collars for inelastic collision studies, and laboratory manual. Table is also equipped with a 1/4-20 threaded hole through its center for circular motion/orbiting experiments.
  1. Details
  2. Images
Fisher HealthCare- Ealing Air Table - 703722
Description Catalog Number Quantity Price
Ealing Air Table
S522293
Science Source
No.:12050
Each for $712.00