Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
S02316
View Image Gallery (1) Image(s) are representative of the product Group
  • S02316
Products for Science Education Fisher Scientific* Timer/Stopwatches
Remarkable timers with large display.
Includes:
  • Magnetic back
  • Flip-open easel stand
  • Spring fastener slip
  • Hole for use with a lanyard
  • Battery
  • Count down from 99 minutes, 59 seconds
  • Alarm can be turned off manually or automatically silenced after one minute
  • Advanced model has a clock and memory
View Specifications
Images for Products for Science Education Fisher Scientific* Timer/Stopwatches
  • S02316
S02316
(2 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Advanced model
S40711 N/AYou must be logged in and have a valid account number to view item availability   Each for $31.00
(0) in My Cart
Basic Model
S02316 N/AYou must be logged in and have a valid account number to view item availability   Each for $44.30
(0) in My Cart
Description
  • Count down from 99 minutes, 59 seconds
  • Alarm can be turned off manually or automatically silenced after one minute
  • Advanced model has a clock and memory
Specifications & Ordering Information:
Dimensions: 6.4L x 5.6W x 1.3cmH; Weight: 1.5 oz. (42.5g)
Certifications: Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA* indicating traceability to NIST*.