Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
14-278-153
New Brunswick Scientific Slit-to-Agar (STA) Air Sampler
Detect airborne microorganism contamination in a variety of environments
·   Microprocessor-controlled, variable-speed turntable and particle distribution guide provide time and duration of contamination event
·   Flowrates and sampling time are entered by using the membrane keypad
·   Sample volume is accurately calculated by measuring and controlling airflow using a thermal mass flow controller; automatically computed and continuously displayed on large three-digit LED
·   Audible and visual alarms signal power interrupt, variation in flowrate, or end of sample run
·   Air volume is drawn by vacuum through a slit orifice and over an exposed agar plate
·   Any contaminants are impacted on the agar surface and after the plate is incubated, the colonies can be counted and identified
View Specifications
Images for New Brunswick Scientific Slit-to-Agar (STA) Air Sampler
  • 14-278-153
14-278-153
14-278-153
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
14-286-121
New Brunswick Scientific
No.:M1075-0100/PRC
Remote Sample Probe for STA-203 Biological Slit-to-Agar Air Samplers
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Description
·   Microprocessor-controlled, variable-speed turntable and particle distribution guide provide time and duration of contamination event
·   Flowrates and sampling time are entered by using the membrane keypad
·   Sample volume is accurately calculated by measuring and controlling airflow using a thermal mass flow controller; automatically computed and continuously displayed on large three-digit LED
·   Audible and visual alarms signal power interrupt, variation in flowrate, or end of sample run
·   Air volume is drawn by vacuum through a slit orifice and over an exposed agar plate
·   Any contaminants are impacted on the agar surface and after the plate is incubated, the colonies can be counted and identified
Compliance: UL, CSA, and CE approved. Requires vacuum pump capable of at least 2.2 cu. ft. air/min. at 15”Hg.
Specifications
ModelSTA-203STA-204
Sample Volume0.03m3 to 5m3 in 0.01m3 increments0.03m3 to 3m3 in 0.01m3 increments
Sample Exposure Time2 to 99.9 min. in 0.1 min. increments2 to 99.9 min. in 0.1 min. increments
Flowrate15 to 50L/min., ±2%15 to 30L/min., ±2%
Electrical Requirements100-240V, 50/60Hz120V, 60Hz
Dimensions (L x W x H)105/8 x 111/4 x 111/2 in. (27 x 28.6 x 29.2cm)105/8 x 111/4 x 18 in. (27 x 28.6 x 45.7cm)
Net Weight11 lb. (5kg)25 lb. (11.4kg)