Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
B1020021
Thermo Scientific™ Shandon™ Downdraft Anatomy Dissecting Table
Two hinged covers totally enclose the work area and the cadaver
Designed to provide a ventilated workstation for animal necropsies, surgeries, perfusions, and dissections. Table functions to pull fumes down and away from the user's face when connected to a negative air source, providing a healthier, safer work environment.
Includes: Removable 20 x 82 in. (50.8 x 208.3cm) body/specimen tray
Construction
  • Type 316 stainless-steel construction
  • Table has two panels, one located at each end of the pan, completely welded in place to direct and balance the exhausting of air within the system
  • Hinged cover includes two positive locks per side to lock the hood in the down position
  • Each hinged cover includes two grab handles that swivel in place to provide ease in lowering and raising the hood
Features
  • Table draws room air equally from both sides via slots on interior side of table perimeter rail, enhancing direction and control of captured air and fumes
  • Pitched table pan allows drainage and collection of waste
  • A sliding viewing port is located on each side of table to allow observation/maintenance of table drain trough below body tray so that tray/specimen does not need to be removed
  • Removable body/specimen tray is pitched to a drain hole and is easily removable for cleaning
  • An adjustable volume damper control is located at end of table for easy access and control of airflow volume across work area
  • An atmospheric damper is included on table to provide continuous airflow volume when table covers are closed, allowing for continuous balance of air on the facility HVAC system
View Specifications
Images for Thermo Scientific™ Shandon™ Downdraft Anatomy Dissecting Table
  • B1020021
B1020021
B1020021
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
23-245-557
Thermo Scientific
No.:B1020021
Downdraft Anatomy Dissection Table
Each for $18,440.00
Check Availability
Add
to Cart
(0) in My Cart
Description
Construction
  • Type 316 stainless-steel construction
  • Table has two panels, one located at each end of the pan, completely welded in place to direct and balance the exhausting of air within the system
  • Hinged cover includes two positive locks per side to lock the hood in the down position
  • Each hinged cover includes two grab handles that swivel in place to provide ease in lowering and raising the hood
Features
  • Table draws room air equally from both sides via slots on interior side of table perimeter rail, enhancing direction and control of captured air and fumes
  • Pitched table pan allows drainage and collection of waste
  • A sliding viewing port is located on each side of table to allow observation/maintenance of table drain trough below body tray so that tray/specimen does not need to be removed
  • Removable body/specimen tray is pitched to a drain hole and is easily removable for cleaning
  • An adjustable volume damper control is located at end of table for easy access and control of airflow volume across work area
  • An atmospheric damper is included on table to provide continuous airflow volume when table covers are closed, allowing for continuous balance of air on the facility HVAC system
Certifications: Agency approvals are available as option; call for details.
Physical Specifications
Dimensions and Weight
Dimensions (D x W x H):32 x 86 x 36 in. (81.3 x 218.4 x 91.4cm)
Weight:350 lb. (158.8kg)
Installation Requirements (customer)
Power RequirementsNone
Plumbing Supply (Waste):Provide one standard 2 in. (5.1cm) copper waste line. If drain trap is required, must be installed by customer.
Ventilation:Call for details.