Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. Tween™ 20, Fisher BioReagents™
Tween™ 20
Viscous, Colorless Liquid
Application: This nonionic detergent is used in the isolation of water-soluble membrane proteins.
Non-ionic detergent .
Polysorbate 20, Polyoxyethylene-20-sorbitan Monolaurate, Polyoxyethylenesorbitan monolaurate
C58H114O26
F.W.: 1227.54
[9005-64-5]
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Tween™ 20, Fisher BioReagents™
  • fisherbiotech
(2 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
BP337-100 Available on GSA/VA Contract for Federal Government customers only.  Glass Bottle; 100mL
MSDS
Each for $36.02
Case of 6 EA for $162.08
Check Availability
Add
to Cart
(0) in My Cart
BP337-500 Available on GSA/VA Contract for Federal Government customers only.  Glass Bottle; 500mL
MSDS
Each for $49.05
Case of 6 EA for $220.70
Check Availability
Add
to Cart
(0) in My Cart
Go to Product Certificates
Product Specifications
Arsenic [As]0.0003% max.
Copper [Cu]0.0005% max.
Lead [Pb]0.0005% max.
IdentificationPass Test
Hydroxyl Value96 to 108
Ignition Residue0.25% max.
ProteaseNot Detected
Water3% max.
Density: 1.11g/mL
Flash Point: 150°C/302°F
Sp. gr.: 1.100
400mPa.s at 25°C
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:0
Recommended storage: RT