Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Pierce GST Protein Interaction Pull-Down Kit, 25-rxn kit
View Image Gallery (3) Image(s) are representative of the product Group
  • Pierce GST Protein Interaction Pull-Down Kit, 25-rxn kit
Thermo Scientific™ Pierce™ GST Protein Interaction Pull-Down Kit
Purify protein interaction complexes with user-supplied GST-tagged fusion protein
Includes: Glutathione agarose resin, lysis buffer, TBS pack, glutathione for elution, microcentrifuge spin columns and collection tubes
Highlights:
  • GST pull-down – purifies protein interactors of any GST-tagged fusion protein
  • Complete kit – provides all components and detailed protocol for purifying protein:protein interactions
  • No special equipment needed – uses common laboratory equipment and reagents (e.g., microcentrifuge)
  • Convenient – microcentrifuge spin columns facilitate simple and efficient manipulation of glutathione agarose beads, including simple processing of multiple samples
  • Flexible – instructions include protocols for bait and prey proteins from different sources
View Specifications
Images for Thermo Scientific™ Pierce™ GST Protein Interaction Pull-Down Kit
  • Pierce GST Protein Interaction Pull-Down Kit, 25-rxn kit
Pierce GST Protein Interaction Pull-Down Kit, 25-rxn kit
Pierce GST Protein Interaction Pull-Down Kit, 25-rxn kit
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
PI-21516
Thermo Scientific
No.:21516
GST Pull-Down Kit; 25-rxn kit
Each for $378.75
Check Availability
Add
to Cart
(0) in My Cart
Description
Highlights:
  • GST pull-down – purifies protein interactors of any GST-tagged fusion protein
  • Complete kit – provides all components and detailed protocol for purifying protein:protein interactions
  • No special equipment needed – uses common laboratory equipment and reagents (e.g., microcentrifuge)
  • Convenient – microcentrifuge spin columns facilitate simple and efficient manipulation of glutathione agarose beads, including simple processing of multiple samples
  • Flexible – instructions include protocols for bait and prey proteins from different sources
Applications: Discover a new protein:protein interaction from a cell lysate; Confirm a putative interaction from a cell lysate or with a previously purified protein; Extract protein:protein interaction information from in vitro transcription/translation lysates