Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
BP2542-10
View Image Gallery (2) Image(s) are representative of the product Group
  • BP2542-10
  • fisherbiotech
Thimerosal, Fisher BioReagents
Thimerosal
White Powder
≥97 %
Sodium ethylmercurithiosalicylate
C9H9HgO2SNa
F.W.: 404.8
[54-64-8]
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
View Specifications
Alert: The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Images for Thimerosal, Fisher BioReagents
  • BP2542-10
  • fisherbiotech
BP2542-10
BP2542-10
(4 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
BP2542-10 Amber Glass; 10g
MSDS
Each for $124.72
Case of 7 EA for $654.80
Check Availability
Add
to Cart
(0) in My Cart
BP2542-100 Amber Glass; 100g
MSDS
Each for $871.40
Case of 3 EA for $2,091.36
Check Availability
Add
to Cart
(0) in My Cart
BP2542-25 Amber Glass; 25g
MSDS
Each for $279.09
Case of 6 EA for $1,255.92
Check Availability
Add
to Cart
(0) in My Cart
BP2542-500 Amber Glass; 500g
MSDS
Each for $2,479.18
Case of 3 EA for $5,950.04
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Product Specifications
pH6.0 to 7.4 (1% Solution)
IdentificationPass Test
Loss on Drying0.5% max.
Melting Point225° to 236°C
Purity97% min.
Decomposition Information: >233°C
Flash Point: 250°C/482°F
Melting Point: 232° to 233°C/449.6° to 451.4°F
DANGER!
Emergency Overview
May be fatal if inhaled, absorbed through skin, or swallowed. Danger of cumulative effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use only under a chemical fume hood. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.
NFPA
Health:4
Flammability:1
Instability:0
Recommended storage: RT, protect from light, desiccate.