Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
14-206-62
View Image Gallery (1) Image(s) are representative of the product Group
  • 14-206-62
Fisherbrand* Absorbent Underpads
Quickly absorb liquids and keep spills from spreading
Cushioned surface minimizes breakage of delicate labware
  • Made with high-absorbency fibers
  • Polyethylene backing is chemical resistant, and the spunbond top layer provides durability even when wet
  • Lie flat, hugging the benchtop to prevent sliding
  • Absorb up to 750mL/m2
View Specifications
Images for Fisherbrand* Absorbent Underpads
  • 14-206-62
14-206-62
(4 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Mat; L x W: 20 x 24 in. (50.8 x 61cm)
14-206-62 N/AYou must be logged in and have a valid account number to view item availability   Pack of 50 for $74.10
Case of 5 PK for $285.50
(0) in My Cart
Mat; L x W: 20 x 36 in.
14-206-63 N/AYou must be logged in and have a valid account number to view item availability   Pack of 25 for $56.50
Case of 5 PK for $220.00
(0) in My Cart
Roll; Dispenser pack; 20 in. x 100 ft.
14-206-64 N/AYou must be logged in and have a valid account number to view item availability   Each for $81.40
Case of 4 EA for $273.00
(0) in My Cart
Roll in dispenser pack; L x W: 20 in. x 250 ft. (51cm x 76m)
14-206-65 N/AYou must be logged in and have a valid account number to view item availability   Each for $150.40
(0) in My Cart
Add Items to Cart
Description
  • Made with high-absorbency fibers
  • Polyethylene backing is chemical resistant, and the spunbond top layer provides durability even when wet
  • Lie flat, hugging the benchtop to prevent sliding
  • Absorb up to 750mL/m2