Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
12-550-15, -17 through -20
Available on GSA/VA Contract for Federal Government customers only. Fisherbrand™ Superfrost™ Plus Microscope Slides
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears
Same outstanding features as regular Superfrost slides with special treatment process that electrostatically adheres tissue sections and cells to the glass without the need for adhesives or protein coatings.
  • Treatment virtually eliminates tissue loss during staining; saves hours of slide preparation
  • Chemical-resistant white marking surface
  • Size: 25 x 75mm
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Fisherbrand™ Superfrost™ Plus Microscope Slides
  • 12-550-15, -17 through -20
12-550-15, -17 through -20
12-550-15, -17 through -20
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
12-550-15 Available on GSA/VA Contract for Federal Government customers only.  Superfrost Plus; White
Pack of 144 for $160.30
Case of 10 PK for $1,314.00
Check Availability
Add
to Cart
(0) in My Cart
22-037-246 Available on GSA/VA Contract for Federal Government customers only.  Superfrost Plus Adhesion
Pack of 144 for $153.09
Pack of 144 for $114.82 Online Instant Savings (25%)
Case of 10 PK for $1,357.38
Case of 10 PK for $1,018.04 Online Instant Savings (25%)
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Treatment virtually eliminates tissue loss during staining; saves hours of slide preparation
  • Chemical-resistant white marking surface
  • Size: 25 x 75mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.