Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
04-355-76
Available on GSA/VA Contract for Federal Government customers only. Decon™ Phenazine pH Test Paper
Packaged in a convenient plastic dispenser with comparative color chart
  • Phenazine paper is designed for determining pH values
  • Provides sharp, clear changes within pH range of 4.5 to 7.5
  • Color scale chart in increments of 0.5pH units
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Decon™ Phenazine pH Test Paper
  • 04-355-76
04-355-76
04-355-76
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
04-355-76 Available on GSA/VA Contract for Federal Government customers only. 
Decon Laboratories
No.:1201
Type: Phenazine; Roll: 0.25 x 180 in. (0.63 x 457cm)
Accessories
Each for $78.66
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Phenazine paper is designed for determining pH values
  • Provides sharp, clear changes within pH range of 4.5 to 7.5
  • Color scale chart in increments of 0.5pH units
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.