Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
19-021-780
Encon Hydrosep Water Treatment Additive for Portable Eyewash Stations
Specifically formulated to provide protection against bacteria, fungi, algae, staph, and acanthamoeba in self-contained eyewash stations. One 8 oz. (237mL) bottle of Hydrosep will protect 5 to 20 gal. (18.9 to 75.7L) of potable water for up to 120 days.
View Specifications
Images for Encon Hydrosep Water Treatment Additive for Portable Eyewash Stations
  • 19-021-780
19-021-780
19-021-780
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
19-021-780
Encon Safety Products
No.:01110764
8 oz.
Case of 4 for $70.38
Check Availability
Add
to Cart
(0) in My Cart
Description
Specifically formulated to provide protection against bacteria, fungi, algae, staph, and acanthamoeba in self-contained eyewash stations. One 8 oz. (237mL) bottle of Hydrosep will protect 5 to 20 gal. (18.9 to 75.7L) of potable water for up to 120 days.
Specifications & Ordering Information:
Supplied four 8 oz. (237mL) bottles to a case.