Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (2) Image(s) are representative of the product Group
  • acros
Available on GSA/VA Contract for Federal Government customers only. Phosphorus pentoxide, 98+%, ACS reagent, ACROS Organics™
Phosphorus pentoxide
ACS reagent
98+%
Application: Adsorbent
Phosphoric anhydride
O5P2
P2O5
F.W.: 141.94
[1314-56-3]
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Phosphorus pentoxide, 98+%, ACS reagent, ACROS Organics™
  • acros
(2 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
  • Special Offer
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
AC315822500
ACROS Organics
No.:315822500
250g; Glass bottle
MSDS
Each for $46.10
Check Availability
Add
to Cart
(0) in My Cart
AC315820010
ACROS Organics
No.:315820010
1kg; Glass bottle
MSDS
Each for $167.90
Check Availability
Add
to Cart
(0) in My Cart
Product Specifications
Assay98.0% min.
Insoluble Matter0.02% max.
Physical FormWhite powder
Ammonium100ppm max.
Heavy Metals [as Pb]100ppm max.
Phosphorus TrioxidePasses Test (limit about 0.02%)
Melting Point: 340.0°C to 360.0°C
Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat.
Sublimation point: 0340.0°C
Danger
GHS P Statement Causes severe skin burns and eye damage. Reacts violently with water.
GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.