Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
21-381 Series
View Image Gallery (2) Image(s) are representative of the product Group
  • 21-381 Series
  • 21-381-116
Eppendorf™ Combitips™ Plus Pipet Tips
Positive displacement for correct volumes
Tips function according to the positive-displacement principle, ensuring that the correct volume is dispensed with regard to the density and viscosity of the liquid used. The positive displacement system provides safer dispensing of radioactive and toxic substances as well as solutions with high vapor pressure. No aerosols are formed.
  • For use with Eppendorf Repeaters™ (original, Plus, Pro, stream and Xstream); and EDOS (automatic dispensing system)
  • Available in nine volumes from 0.1 to 50mL
  • Reliable results ensured by positive-displacement system
  • Fine tip of Combitip provides exact dispensing without additional pipet tip
  • Polyethylene plunger and polypropylene cylinder make these tips ideal for use with aggressive reagents
  • Choose from sterile and nonsterile Combitips Plus Tips and Eppendorf Biopur™ tips, which are certified sterile as well as pyrogen-, RNase-, DNA-, and ATP-free
View Specifications
Images for Eppendorf™ Combitips™ Plus Pipet Tips
  • 21-381 Series
  • 21-381-116
21-381 Series
21-381 Series
(23 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
21-381-341
Eppendorf North America Biotools
No.:022267701
Sterile; Volume: 50mL; Dispensing range: 500µL to 10mL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-326
Eppendorf North America Biotools
No.:022266004
Nonsterile; Volume: 0.2mL; Dispensing range: 2 to 40µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-334
Eppendorf North America Biotools
No.:022267001
Sterile; Volume: 0.1mL; Dispensing range: 1 to 20µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-335
Eppendorf North America Biotools
No.:022267108
Sterile; Volume: 0.2mL; Dispensing range: 2 to 40µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-336
Eppendorf North America Biotools
No.:022267205
Sterile; Volume: 0.5mL; Dispensing range: 5 to 100µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-403
Eppendorf North America Biotools
No.:022496069
Eppendorf Biopur; Volume: 1.0mL; Dispensing range: 10 to 200µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-405
Eppendorf North America Biotools
No.:022496107
Eppendorf Biopur; Volume: 5.0mL; Dispensing range: 50 to 1000µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-404
Eppendorf North America Biotools
No.:022496085
Eppendorf Biopur; Volume: 2.5mL; Dispensing range: 25 to 500µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-407
Eppendorf North America Biotools
No.:022496140
Eppendorf Biopur; Volume: 50mL; Dispensing range:500µL to 10mL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-400
Eppendorf North America Biotools
No.:022496000
Eppendorf Biopur; Volume: 0.1mL; Dispensing range: 1 to 20µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-340
Eppendorf North America Biotools
No.:022267604
Sterile; Volume: 10mL; Dispensing range: 100 to 2000uL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-115
Eppendorf North America Biotools
No.:022266551
Nonsterile; Volume: 25mL; Dispensing range: 250µL to 5mL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-329
Eppendorf North America Biotools
No.:022266306
Nonsterile; Volume: 2.5mL; Dispensing range: 25 to 500uL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-327
Eppendorf North America Biotools
No.:022266101
Nonsterile; Volume: 0.5mL; Dispensing range: 5 to 100µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-331
Eppendorf North America Biotools
No.:022266501
Nonsterile; Volume: 10mL; Dispensing range: 100 to 2000uL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-116
Eppendorf North America Biotools
No.:022266624
Assortment Pack (5 each size plus one 50mL, one 25mL adapter); Used w/Eppendorf Repeater Plus
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
21-381-402
Eppendorf North America Biotools
No.:022496042
Eppendorf Biopur; Volume: 0.5mL; Dispensing range: 5 to 100µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-406
Eppendorf North America Biotools
No.:022496123
Sterile; Volume: 10mL; Dispensing range: 100 to 2000µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-339
Eppendorf North America Biotools
No.:022267507
Sterile; Volume: 5.0mL; Dispensing range: 50 to 1000µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-328
Eppendorf North America Biotools
No.:022266209
Nonsterile; Volume: 1.0mL; Dispensing range: 10 to 200µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-325
Eppendorf North America Biotools
No.:022265954
Nonsterile; Volume: 0.1mL; Dispensing range: 1 to 20µL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-330
Eppendorf North America Biotools
No.:022266403
Nonsterile; Volume: 5.0mL; Dispensing range: 50 to 1000uL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-381-332
Eppendorf North America Biotools
No.:022266608
Nonsterile; Volume: 50mL; Dispensing range: 500µL to 10mL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
  • For use with Eppendorf Repeaters™ (original, Plus, Pro, stream and Xstream); and EDOS (automatic dispensing system)
  • Available in nine volumes from 0.1 to 50mL
  • Reliable results ensured by positive-displacement system
  • Fine tip of Combitip provides exact dispensing without additional pipet tip
  • Polyethylene plunger and polypropylene cylinder make these tips ideal for use with aggressive reagents
  • Choose from sterile and nonsterile Combitips Plus Tips and Eppendorf Biopur™ tips, which are certified sterile as well as pyrogen-, RNase-, DNA-, and ATP-free
Performance Data for Combitip Plus Tip†
Tip CapacityTest VolumeInaccuracyImprecision
For Repeater Plus
0.1mL2µL±1.6%<=3.0%
 20µL±1.0%<=2.0%
0.2mL4µL±1.3%<=2.0%
 40µL±0.8%<=1.5%
0.5mL10µL±0.9%<=1.5%
 100µL±0.8%<=0.6%
1.0mL20µL±0.9%<=0.9%
 200µL±0.6%<=0.4%
2.5mL50µL±0.8%<=0.8%
 500µL±0.5%<=0.3%
5.0mL100µL±0.6%<=0.6%
 1000µL±0.5%<=0.25%
10mL200µL±0.5%<=0.6%
 2000µL±0.5%<=0.25%
25mL500µL±0.4%<=0.6%
 5000µL±0.3%<=0.25%
50mL1000µL±0.3%<=0.5%
 10L±0.3%<=0.25%
For Repeater stream and Xstream
0.1mL10µL±1.6%<=2.5%
 100µL±1.0%<=0.5%
0.2mL20µL±1.3%<=1.5%
 200µL±1.0%<=0.5%
0.5mL50µL±0.9%<=0.8%
 500µL±0.9%<=0.3%
1mL100µL±0.9%<=0.55%
 1000µL±0.6%<=0.2%
2.5mL250µL±0.8%<=0.45%
 2500µL±0.5%<=0.15%
5mL500µL±0.8%<=0.35%
 5000µL±0.5%<=0.15%
10mL1000µL±0.5%<=0.25%
 10,000µL±0.4%<=0.15%
25mL2500µL±0.3%<=0.25%
 25,000µL±0.3%<=0.15%
50mL5000µL±0.3%<=0.25%
† Bidistilled water at 20° to 25°C; constant to ±0.5°C. Number of determinations: 15 according to DIN 12 650.