Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
AC24164-0000
View Image Gallery (2) Image(s) are representative of the product Group
  • AC24164-0000
  • acros
Silica gel, for drying purposes, blue indicating, 2-5mm, ACROS Organics™
Silica gel
Application: Drying agent, for column chromatography, Indicator
O2Si
F.W.: 60.08
[7631-86-9], [7646-79-9]
View Specifications
Images for Silica gel, for drying purposes, blue indicating, 2-5mm, ACROS Organics™
  • AC24164-0000
  • acros
AC24164-0000
AC24164-0000
(2 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
  • Special Offer
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
AC24164-0010
ACROS Organics
No.:241640010
1kg
MSDS
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
AC24164-0050
ACROS Organics
No.:241640050
5kg
MSDS
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Product Specifications
pH2.5 to 6.5
Physical FormBlue granules
Bulk Density0.75 to 0.95
Loss on Drying1% max. (160°C)
Particle Size<2mm: 0 to 7%, <5mm: 93 to 100%
Water Soluble Substancessalts soluble in water: 0.8 to 2.6%
Water Adsorption> 18% (25°C, relative humidity 40%)
Melting Point: 160°C
Solubility in water: insoluble
MOISTURE SENSITIVE
GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May cause cancer.
Toxic if inhaled.
May cause cancer by inhalation.
May damage fertility.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.