Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. Magnesium Oxide (Heavy Powder/USP/FCC), Fisher Chemical
Magnesium Oxide
Heavy Powder
USP/FCC
MgO
F.W.: 40.3
[1309-48-4]
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Magnesium Oxide (Heavy Powder/USP/FCC), Fisher Chemical
  • fisherchemical
(2 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
M68-3 Available on GSA/VA Contract for Federal Government customers only.  Poly Pail; 3kg
MSDS
Each for $327.75
Check Availability
Add
to Cart
(0) in My Cart
M68-500 Poly Bottle; 500g
MSDS
Each for $224.12
Case of 4 EA for $678.45
Check Availability
Add
to Cart
(0) in My Cart
Go to Product Certificates
Product Specifications
Arsenic [As]3mg/kg max. (FCC)
Assay96.0 to 100.5% (FCC/USP)
Calcium [Ca]1.1% max. (USP)
Iron [Fe]0.05% max. (USP)
Lead [Pb]10mg/kg max. (FCC)
Insoluble in Acid0.1% max. (FCC/USP)
Free Alkali and Soluble Salts2.0% max. (USP)
Bulk DensityReport g/mL (USP)
Calcium Oxide1.5% max. (FCC)
Heavy Metals [as Pb]20µg/g max. (USP)
IdentificationPass Test (FCC/USP)
Loss on Ignition10.0% max. (FCC/USP)
Residual SolventsMeets Requirements (USP)
Boiling Point: 3600°C
Density: 3.58g/mL
Melting Point: 2800°C
Signal Word: CAUTION!
Emergency Overview
May cause eye, skin and respiratory tract irritation.
NFPA
Health: 1
Flammability: 0
Instability: 1