Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
EMD Millipore Fluids Contamination Analysis Kit
Conforms to standard contamination test procedures
Includes: Glass filter holder (47mm) with stainless steel screen, 1L Vacuum filtering flask, blunt end filter forceps, latex tubing (1/4 in. x 23 in.), 25mm Solvent Filtering Dispenser, 400 PetriSlides, Chemical Duty Pump (115V), Chemical Duty Pump (220V).
  • Conforms to ARP-598 (hydraulic fluids), ASTM-D2276 (aviation turbine fuels), and federal standard 791a (lubricants, liquid fuels, and related products)
  • Includes expendable materials for approximately 600 tests
View Specifications
Images for EMD Millipore Fluids Contamination Analysis Kit
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
XX71 047 12
EMD Millipore
No.:XX7104712
Fluids Contamination Analysis Kit; 230V/50Hz
Accessories
Each for $2,150.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Conforms to ARP-598 (hydraulic fluids), ASTM-D2276 (aviation turbine fuels), and federal standard 791a (lubricants, liquid fuels, and related products)
  • Includes expendable materials for approximately 600 tests