Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Pentra™ 60 C+ (23-450-042)
HORIBA ABX Pentra™ 60 C + Hematology Analyzer
Five-part leukocyte differential analysis through a combination of cytochemistry, light absorbency technology, and focused-flow volumetric determination with selectable CBC or CBC plus five-part differential analysis
Proven, innovative technology for ABX Pentra™ 60 C+
DHSS™ (Double Hydrodynamic Sequential System)
  • Allows measurement of cell volumes and cellular content in one unique chamber
  • Following the cell incubation with Eosinofix™, a cytochemical reagent containing the active principle of Sudan Black B leukocytes is analyzed according to two complementary principles: focused-flow impedance (cell volume measurement) and light absorbance (cellular content analysis)
  • Offers specific advantages for:
  • Post-draw stability of the leukocyte differential analysis for up to 48 hours
  • High resolution leukocyte differential
  • Identification and counting of two additional leukocyte subsets: Large Immature Cells (LIC) and Atypical Lymphocytes (ALY)
MDSS™ (Multi Distribution Sampling System)
  • Consists in aspirating a volume of 53µL of whole blood in the sample needle
  • This volume of blood is then split in different aliquots within the sampling needle and distributed in all specific chambers for the appropriate dilutions
  • Aliquots of blood are distributed in a tangential flow of diluent
  • Needle is positioned near the chamber cavity and injects the aliquot of blood in the reagent tangential flow
  • Sampling needle is then cleaned internally and externally
  • Eliminates use of shear-valve to distribute the blood sample in all appropriate chambers and does not require any maintenance
  • The ABX patented tangential flow technique gives a perfect mixing and homogenization of the blood with the reagents
  • Distribution of blood in a tangential flow eliminates viscosity problems coming from the whole blood sample
  • Also gives the possibility of immediate CBC/5 DIFF test selection without a cleaning cycle in between
View Specifications
Images for HORIBA ABX Pentra™ 60 C + Hematology Analyzer
  • Pentra™ 60 C+ (23-450-042)
Pentra™ 60 C+ (23-450-042)
Pentra™ 60 C+ (23-450-042)
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
23-450-042
ABX Diagnostics
No.:P60CP001NUA1
Closed tube; 26 parameters
Description
Proven, innovative technology for ABX Pentra™ 60 C+
DHSS™ (Double Hydrodynamic Sequential System)
  • Allows measurement of cell volumes and cellular content in one unique chamber
  • Following the cell incubation with Eosinofix™, a cytochemical reagent containing the active principle of Sudan Black B leukocytes is analyzed according to two complementary principles: focused-flow impedance (cell volume measurement) and light absorbance (cellular content analysis)
  • Offers specific advantages for:
  • Post-draw stability of the leukocyte differential analysis for up to 48 hours
  • High resolution leukocyte differential
  • Identification and counting of two additional leukocyte subsets: Large Immature Cells (LIC) and Atypical Lymphocytes (ALY)
MDSS™ (Multi Distribution Sampling System)
  • Consists in aspirating a volume of 53µL of whole blood in the sample needle
  • This volume of blood is then split in different aliquots within the sampling needle and distributed in all specific chambers for the appropriate dilutions
  • Aliquots of blood are distributed in a tangential flow of diluent
  • Needle is positioned near the chamber cavity and injects the aliquot of blood in the reagent tangential flow
  • Sampling needle is then cleaned internally and externally
  • Eliminates use of shear-valve to distribute the blood sample in all appropriate chambers and does not require any maintenance
  • The ABX patented tangential flow technique gives a perfect mixing and homogenization of the blood with the reagents
  • Distribution of blood in a tangential flow eliminates viscosity problems coming from the whole blood sample
  • Also gives the possibility of immediate CBC/5 DIFF test selection without a cleaning cycle in between
  • 26 parameters; 60 samples/hour
  • Full, 5-part differential
  • “Zero-maintenance” concept
  • Five-part leukocyte differential analysis through a combination of cytochemistry, light absorbency technology, and focused-flow volumetric determination with selectable CBC or CBC plus five-diff analysis
  • Offers a “hands-free” system for either open- or closed-tube sampling
  • Requiring only a 53µL sampling volume, multiple aspirations are possible from fingerstick collection devices
  • QC, Calibration, Patient Data Management, and all instrument functions are handled via the all-in-one, “low profile” data manager (for PENTRA 60 C+ only), complete with a 15 in. flat screen monitor
  • Retractable probe is washed internally and externally to minimize biohazard exposure to the sample, improving precision and reducing carryover
Proven, innovative technology for ABX Pentra™ 60 C+
DHSS™ (Double Hydrodynamic Sequential System)
  • Allows measurement of cell volumes and cellular content in one unique chamber
  • Following the cell incubation with Eosinofix™, a cytochemical reagent containing the active principle of Sudan Black B leukocytes is analyzed according to two complementary principles: focused-flow impedance (cell volume measurement) and light absorbance (cellular content analysis)
  • Offers specific advantages for:
  • Post-draw stability of the leukocyte differential analysis for up to 48 hours
  • High resolution leukocyte differential
  • Identification and counting of two additional leukocyte subsets: Large Immature Cells (LIC) and Atypical Lymphocytes (ALY)
MDSS™ (Multi Distribution Sampling System)
  • Consists in aspirating a volume of 53µL of whole blood in the sample needle
  • This volume of blood is then split in different aliquots within the sampling needle and distributed in all specific chambers for the appropriate dilutions
  • Aliquots of blood are distributed in a tangential flow of diluent
  • Needle is positioned near the chamber cavity and injects the aliquot of blood in the reagent tangential flow
  • Sampling needle is then cleaned internally and externally
  • Eliminates use of shear-valve to distribute the blood sample in all appropriate chambers and does not require any maintenance
  • The ABX patented tangential flow technique gives a perfect mixing and homogenization of the blood with the reagents
  • Distribution of blood in a tangential flow eliminates viscosity problems coming from the whole blood sample
  • Also gives the possibility of immediate CBC/5 DIFF test selection without a cleaning cycle in between
PENTRA 60 Series Specifications
CBC Analysis (12 parameters)
WBC; RBC; Hgb; Hct; MCV; MCH; MCHC; RDW; Pct; MPV; PLT
CBC plus 5 DIFF Analysis (26 parameters)
WBC; LYM2; MON2; NEU2; EOS; BAS2; LIC1,2; ALY1,2; RBC; Hgb; Hct; MCV; MCH; MCHC; RDW; PDW1; Pct; MPV; PLT
Technical Specifications
StabilityUp to 48 hr.
Throughput60 samples/hr.
Sample Volume30 µL (CBC); 53 µL (5 diff)
Sample I.D.Alphanumeric keyboard; Bar code reader
Reagents5
PrinterDot matrix (PENTRA 60); Laser (PENTRA 60 C+)
Dimensions17.5 in.W x 15 in.D x 22.3 in.H
Weight77 lbs.
Electrical Requirements110/220V; 50/60 Hz; 200VA
Data Management (PENTRA 60 C+)PC (1.8GHZ); RAM: 256 Mb; Hard Drive: 40 Gb; Floppy disk; CD-Rom; 15 in. screen (600 x 800)
1For research only.
2In percentage (%) and absolute value (#).
For research use.