Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Extraction Disks
3M™ Empore™ C8 and C18 Extraction Disks
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
View Specifications
Images for 3M™ Empore™ C8 and C18 Extraction Disks
  • Extraction Disks
Extraction Disks
Extraction Disks
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
14-386
3M Empore
No.:2214
C8 Sorbent; 47mm
14-386-2
3M Empore
No.:2215
C18 Sorbent; 47mm
14-386-3
3M Empore
No.:2315
C18 Sorbent; 90mm
14-378-061
3M Empore
No.:2314
C8 Sorbent; 90mm
Description
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
  • Effective for extracting PCBs, PNAs, organochlorine pesticides, phenoxy acid herbicides, phthalates, and triazines
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
  • C8 disk is approved for use in EPA Method 549.1
  • C18 disks are approved for use in EPA Methods 506, 550.1, 525.2, 1613B, 508.1, and 3535; 14-386-3 is also approved for use in EPA Method 608