Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
08-215-2
CoorsTek™ Porcelain Rose Crucible
Withstands temperatures up to 1400°C (2550°F), not to exceed 200°C/hour
  • Unglazed porcelain crucible
  • Eliminates possibility of reaction between contents and glaze coating
View Specifications
Images for CoorsTek™ Porcelain Rose Crucible
  • 08-215-2
08-215-2
08-215-2
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
08-215-2
CoorsTek
No.:60166
75mL
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
  • Unglazed porcelain crucible
  • Eliminates possibility of reaction between contents and glaze coating